Skip to Content
Merck
All Photos(1)

Key Documents

40263

Sigma-Aldrich

N,N-Dimethylformamide di-tert-butyl acetal

technical, ≥90% (GC)

Synonym(s):

1,1-Di-tert-butoxy-N,N-dimethylmethylamine, 1,1-Di-tert-butoxytrimethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH[OC(CH3)3]2
CAS Number:
Molecular Weight:
203.32
Beilstein:
969629
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

Assay

≥90% (GC)

form

liquid

refractive index

n20/D 1.413 (lit.)

bp

56-57 °C/8 mmHg (lit.)

density

0.848 g/mL at 25 °C (lit.)

functional group

amine
ether

SMILES string

CN(C)C(OC(C)(C)C)OC(C)(C)C

InChI

1S/C11H25NO2/c1-10(2,3)13-9(12(7)8)14-11(4,5)6/h9H,1-8H3

InChI key

DBNQIOANXZVWIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N-Dimethylformamide di-tert-butyl acetal may be used in the synthesis of following:
  • (S)-2-(1-tert-butoxycarbonyl-2-{4-[2-(5-methyl-2-phenyloxazol-4-yl)ethoxy]phenyl}ethylamino)benzoic acid methyl ester
  • 3-O-tert-butylmorphine
  • tert-butylesters of pyrrole- and indolecarboxylic acids

Other Notes

Review; synthesis of tert-butyl esters.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Mohacsi et al.
Journal of medicinal chemistry, 25(10), 1264-1266 (1982-10-01)
3-O-tert-Butylmorphine (5) was prepared from 6-O-acetylmorphine (3) via alkylation with N,N-dimethylformamide di-tert-butyl acetal, followed by hydrolytic removal of the 3-(dimethylamino)-2-propenoate group. The same process was used to prepare the tert-butyl ether of levorphanol (6), (-)-3-tert-butoxy-N-methylmorphinan (8). Both 5 and 8
Convenient synthesis of pyrrole-and indolecarboxylic acid tert-butylesters.
Ludwig J and Lehr M.
Synthetic Communications, 34(20), 3691-3695 (2004)
William B Mathews et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 46(10), 1719-1726 (2005-10-06)
The goal of this study was to synthesize and evaluate in vivo the peroxisome proliferator-activated receptor-gamma (PPARgamma) agonist (11)C-GW7845 ((S)-2-(1-carboxy-2-{4-[2-(5-methyl-2-phenyloxazol-4-yl)ethoxy]phenyl}ethylamino)benzoic acid methyl ester) ((11)C-compound 1). PPARgamma is a member of a family of nuclear receptors that plays a central role
U. Widmer
Synthesis, 135-135 (1983)
R.F. Abdulla et al.
Tetrahedron, 35, 1675-1675 (1979)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service