Recommended Products
Assay
98%
form
liquid
refractive index
n20/D 1.478 (lit.)
bp
148-149 °C (lit.)
density
1.425 g/mL at 25 °C (lit.)
functional group
chloro
storage temp.
2-8°C
SMILES string
COC(=N)C(Cl)(Cl)Cl
InChI
1S/C3H4Cl3NO/c1-8-2(7)3(4,5)6/h7H,1H3
InChI key
OGBINJLTBZWRRB-UHFFFAOYSA-N
Related Categories
Application
Methyl 2,2,2-trichloroacetimidate was used as starting material during the synthesis of novel bibenzimidazole oligomers and polymers. It was employed as reagent during the synthesis of 2,2′-bisbenzimidazole-5,5′-dicarboxylic acid.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
165.2 °F - closed cup
Flash Point(C)
74 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
A new sequence isomer of AB-polybenzimidazole for high-temperature PEM fuel cells.
Journal of Polymer Science Part A: Polymer Chemistry, 50(2), 306-313 (2012)
The Journal of organic chemistry, 70(23), 9436-9446 (2005-11-05)
[Structure: see text]. A simple and mild condensation route for the synthesis of novel bibenzimidazole oligomers and polymers is reported here using methyl 2,2,2-trichloroacetimidate as a key starting material. The dimer, trimer, tetramer, and polymers of bibenzimidazole were synthesized as
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service