Skip to Content
Merck
All Photos(1)

Key Documents

13929

Sigma-Aldrich

Benzyl thiocyanate

≥95.0% (GC)

Synonym(s):

Benzyl rhodanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2SCN
CAS Number:
Molecular Weight:
149.21
Beilstein:
1859726
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (GC)

form

solid

bp

230-235 °C (lit.)

mp

39-41 °C (lit.)
39-41 °C

solubility

diethyl ether: soluble 0.5 g/10 mL, clear to very faintly turbid, colorless to almost colorless

functional group

phenyl
thiocyanate
thioether

storage temp.

2-8°C

SMILES string

N#CSCc1ccccc1

InChI

1S/C8H7NS/c9-7-10-6-8-4-2-1-3-5-8/h1-5H,6H2

InChI key

ABNDFSOIUFLJAH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzyl thiocyanate undergoes regioselective bond dissociation during its electrochemical reduction in acetonitrile at an inert electrode. It is added to stimulate the chlortetracycline biosynthesis during industrial fermentations. It undergoes biotransformation into dibenzyl disulphide by Streptomyces aureofaciens.

Application

Benzyl thiocyanate was used to study the effects of various dietary compounds on the α-hydroxylation of N-nitrosopyrrolidine in male F344 rats in vitro.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Obstructive cholangiopathy and isothiocyanate].
D W Bleyl et al.
Die Nahrung, 35(7), 783-785 (1991-01-01)
S Sugie et al.
Japanese journal of cancer research : Gann, 84(8), 865-870 (1993-08-01)
The effects of two aromatic thiocyanates, benzyl isothiocyanate (BITC) and benzyl thiocyanate (BTC), on diethylnitrosamine (DEN)-induced hepatocarcinogenesis were examined in rats. A total of 108 male ACI/N rats, 5 weeks old, were divided into 6 groups (18 rats in each).
J Novotná et al.
Current microbiology, 31(2), 84-91 (1995-08-01)
Cell protein profiles of submerged cultures of Streptomyces aureofaciens cultivated in the absence or presence of 12 microM benzyl thiocyanate (BT) were analyzed by one-dimensional SDS polyacrylamide gel electrophoresis. Substantial increase in the intensity of the 13, 35, 37, 60
S Sugie et al.
Carcinogenesis, 14(2), 281-283 (1993-02-01)
The effects of benzyl isothiocyanate (BITC) and benzyl thiocyanate (BTC) on two types of DNA synthesis were examined in hepatocyte primary cultures (HPC). Male F344 rats were fed BITC- or BTC-containing diets at a concentration of 400 p.p.m. Using hepatocytes
V Erban et al.
Folia microbiologica, 32(5), 411-416 (1987-01-01)
The localization of anhydrotetracycline oxygenase and glucose-6-phosphate dehydrogenase (EC 1.1.1.49) was studied by determining the enzyme activities in subcellular fractions obtained by differential centrifugation of the mycelia of Streptomyces aureofaciens after lysozyme treatment. Glucose-6-phosphate dehydrogenase was a typical cytoplasmic enzyme

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service