Skip to Content
Merck
All Photos(2)

Key Documents

139254

Sigma-Aldrich

2,4-Dichlorobenzyl chloride

99%

Synonym(s):

α,2,4-Trichlorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3CH2Cl
CAS Number:
Molecular Weight:
195.47
Beilstein:
387220
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.576 (lit.)

bp

248 °C (lit.)

mp

−2.6 °C (lit.)

density

1.407 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

ClCc1ccc(Cl)cc1Cl

InChI

1S/C7H5Cl3/c8-4-5-1-2-6(9)3-7(5)10/h1-3H,4H2

InChI key

IRSVDHPYXFLLDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,4-Dichlorobenzyl chloride was used as starting reagent for the synthesis of series of novel 1,2,4-triazolium derivatives[1].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Luo et al.
Archiv der Pharmazie, 342(7), 386-393 (2009-06-23)
A series of novel 1,2,4-triazolium derivatives was synthesized starting from commercially available 1H-1,2,4-triazole, 2,4-dichlorobenzyl chloride, or 2,4-difluorobenzyl bromide. Their antibacterial and antifungal activities were evaluated against Staphylococcus aureus ATCC 29213, Escherichia coli ATCC 25922, Bacillus proteus, Bacillus subtilis, Pseudomonas aeruginosa

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service