Skip to Content
Merck
All Photos(1)

Key Documents

13609

Sigma-Aldrich

Wang resin

100-200 mesh particle size, extent of labeling: 1.1 mmol/g loading

Synonym(s):

p-Alkoxy-benzyl alcohol polymer-bound, p-Alkoxybenzyl alcohol resin, [4-(Hydroxymethyl)phenoxymethyl]polystyrene

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

extent of labeling

1.1 mmol/g loading

matrix

crosslinked with 1% DVB

particle size

100-200 mesh

application(s)

peptide synthesis

functional group

alcohol

Looking for similar products? Visit Product Comparison Guide

General description

Wang resin also known as p-Alkoxybenzyl alcohol resin, is commonly used as a solid support in the synthesis of peptides.

Application

Wang resin is used as a solid support to obtain cyclotides via solid phase peptide synthesizing method.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of Kalata B2 cyclotide (GLPVCGETCFGGTCNTPGCSCTWPICTRD) on wang resin, as solid support
V Deepshikha
Open Journal of Medicinal Chemistry, 10, 46-55 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service