Skip to Content
Merck
All Photos(1)

Key Documents

8.55011

Sigma-Aldrich

Merrifield resin HL (100-200 mesh)

Novabiochem®

Synonym(s):

Chloromethylpolystyrene-divinylbenzene (100-200 mesh)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352101
NACRES:
NA.22

Pricing and availability is not currently available.

Quality Level

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Boc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

2-30°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
Z304832Z304824SYNA871922
manufacturer/tradename

Quickfit MF 23/3

manufacturer/tradename

Quickfit MF 23/2

manufacturer/tradename

Quickfit MF 23/1

manufacturer/tradename

Synthware A871922

binding type

non-treated surface

binding type

non-treated surface

binding type

non-treated surface

binding type

-

material

colorless tube, glass tube

material

colorless tube, glass tube

material

colorless tube, glass tube

material

colorless tube, glass tube

joint

joint: ST/NS24/29

joint

joint: ST/NS19/26

joint

joint: ST/NS14/23

joint

joint: ST/NS19/22

feature

graduations: no

feature

graduations: no, neck thread PUSH FIT (19/26 cone size)

feature

graduations: no, neck thread PUSH FIT (14/23 cone size)

feature

-

General description

High load version of standard chloromethylpolystyrene.

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Linkage

Replaces: 01-64-0070

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (determined by elemental analysis for sulfur after derivatization with KSAc): 1.0 - 1.6 mmol/g
Swelling Volume (in DMF): lot specific result
The polymer matrix is copoly (styrene-1 % DVB), 100 -200 mesh

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Anhydrous HF is the preferred reagent for peptide cleavage from Boc-based resins, versatile and effective for various peptide synthesis.

Anhydrous HF is the preferred reagent for peptide cleavage from Boc-based resins, versatile and effective for various peptide synthesis.

Anhydrous HF is the preferred reagent for peptide cleavage from Boc-based resins, versatile and effective for various peptide synthesis.

Anhydrous HF is the preferred reagent for peptide cleavage from Boc-based resins, versatile and effective for various peptide synthesis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service