134317
trans-2-Phenylcyclopropyl isocyanate
90%, remainder predominantly cis
About This Item
Recommended Products
Assay
90%
form
liquid
refractive index
n20/D 1.546 (lit.)
bp
75-76 °C/0.5 mmHg (lit.)
density
1.074 g/mL at 25 °C (lit.)
functional group
isocyanate
SMILES string
O=C=N[C@@H]1C[C@H]1c2ccccc2
InChI
1S/C10H9NO/c12-7-11-10-6-9(10)8-4-2-1-3-5-8/h1-5,9-10H,6H2/t9-,10+/m0/s1
InChI key
DYUXVJAFBUZREW-VHSXEESVSA-N
Application
accessory
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
No data available
Flash Point(C)
No data available
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service