Skip to Content
Merck
All Photos(3)

Key Documents

120979

Sigma-Aldrich

2,3,5-Triiodobenzoic acid

98%

Synonym(s):

TIBA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
I3C6H2CO2H
CAS Number:
Molecular Weight:
499.81
Beilstein:
1955088
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

220-222 °C (lit.)

solubility

methanol: soluble 5%

functional group

carboxylic acid

SMILES string

OC(=O)c1cc(I)cc(I)c1I

InChI

1S/C7H3I3O2/c8-3-1-4(7(11)12)6(10)5(9)2-3/h1-2H,(H,11,12)

InChI key

ZMZGFLUUZLELNE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,3,5-Triiodobenzoic acid has been used in the synthesis of aromatic iodine containing vinylic monomer, 2-methacryloyloxyethyl(2,3,5-triiodobenzoate) used in preparing radiopaque polymers.

Biochem/physiol Actions

2,3,5-Triiodobenzoic acid (TIBA) inhibits the translocation of indole-3-acetic acid transport.TIBA inhibits the colonization of the main root cortex by Laccaria bicolor S238 N and the formation of the Hartig net.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Anna Galperin et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 83(2), 490-498 (2007-04-28)
Radiopaque magnetic gamma-Fe(2)O(3)/poly(2-methacryloyloxyethyl(2,3,5-triiodobenzoate)) core-shell nanoparticles of narrow size distribution were prepared by emulsion polymerization of the iodinated monomer 2-methacryloyloxyethyl(2,3,5-triiodobenzoate) in the presence of maghemite (gamma-Fe(2)O(3) nanoparticles coated with a dextran shell are commonly used as contrast agents for magnetic resonance
The Effect of 2,4-Dichlorophenoxyacetic Acid and 2,3,5-Triiodobenzoic Acid on the Transport of Indoleacetic Acid.
J R Hay
Plant physiology, 31(2), 118-120 (1956-03-01)
The auxin transport inhibitor 2, 3, 5-triiodobenzoic acid (TIBA) inhibits the stimulation of in vitro lateral root formation and the colonization of the tap-root cortex of Norway spruce (Picea abies) seedlings by the ectomycorrhizal fungus Laccaria bicolor.
KARABAGHLI-DEGRON C, et al.
The New phytologist, 140(4), 723-733 (1998)
H G Mahesha et al.
The FEBS journal, 273(3), 451-467 (2006-01-20)
Genistein and daidzein, the major isoflavones present in soybeans, possess a wide spectrum of physiological and pharmacological functions. The binding of genistein to human serum albumin (HSA) has been investigated by equilibrium dialysis, fluorescence measurements, CD and molecular visualization. One
Li Dong et al.
Plant molecular biology, 60(4), 599-615 (2006-03-10)
Previously, we characterized 92 Arabidopsis genes (AtSFLs) similar to the S-locus F-box genes involved in S-RNase-based self-incompatibility and found that they likely play diverse roles in Arabidopsis. In this study, we investigated the role of one of these genes, CEGENDUO

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service