Skip to Content
Merck
All Photos(1)

Key Documents

133329

Sigma-Aldrich

Formanilide

99%

Synonym(s):

N-Phenylformamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHCHO
CAS Number:
Molecular Weight:
121.14
Beilstein:
906934
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

powder

bp

166 °C/14 mmHg (lit.)

mp

46-48 °C (lit.)

solubility

water: soluble 25.4 g/L at 20 °C
water: soluble 28.6 g/L at 25 °C

density

1.144 g/mL at 25 °C (lit.)

SMILES string

O=CNc1ccccc1

InChI

1S/C7H7NO/c9-6-8-7-4-2-1-3-5-7/h1-6H,(H,8,9)

InChI key

DYDNPESBYVVLBO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Formanilide was used to study the zero electron kinetic energy(ZEKE) spectra of cis- and trans-formanilide. It was used to investigate the gas-phase structures of the two isomers of the trans-formanilide-water complex by two-colour (1+1′) resonance enhanced multiphoton ionisation (REMPI) and ZEKE spectroscopy.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydration of a cationic amide group: a ZEKE spectroscopic study of trans-formanilide-H2O.
Ullrich S, et al.
Physical Chemistry Chemical Physics, 4(13), 2897-2903 (2002)
ZEKE photoelectron spectroscopy of the cis and trans isomers of formanilide.
Susanne Ullrich et al.
Angewandte Chemie (International ed. in English), 41(1), 166-168 (2002-12-20)
Aminolysis and hydrolysis of formanilide in water solutions. V. Influence of the substituent in para-position.
B Bergstrand
Acta pharmaceutica Suecica, 22(1), 1-16 (1985-01-01)
Mitsuhiko Miyazaki et al.
Physical chemistry chemical physics : PCCP, 11(29), 6098-6106 (2009-07-17)
We measured the infrared (IR) spectra of supersonically cooled N-phenylformamide (formanilide) and N-phenylacetamide (acetanilide) in the amide band and X-H stretch vibration regions by using IR-UV depletion spectroscopy combined with a newly developed mid-IR light source based on difference frequency
Johnson K Agbo et al.
The Journal of chemical physics, 127(6), 064315-064315 (2007-08-21)
A potential energy surface for trans-formanilide (TFA)-H2O is calculated and applied to study energy flow in the complex as well as the kinetics of water shuttling between hydrogen bonding sites on TFA. In addition to the previously identified H2O-TFA(C[Double Bond]O)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service