Skip to Content
Merck
All Photos(1)

Key Documents

131482

Sigma-Aldrich

4-Phenylthiosemicarbazide

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHCSNHNH2
CAS Number:
Molecular Weight:
167.23
Beilstein:
608285
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

138-140 °C (lit.)

functional group

amine

SMILES string

NNC(=S)Nc1ccccc1

InChI

1S/C7H9N3S/c8-10-7(11)9-6-4-2-1-3-5-6/h1-5H,8H2,(H2,9,10,11)

InChI key

KKIGUVBJOHCXSP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Phenylthiosemicarbazide was used in the synthesis of amberlite XAD resins. It was also used in the synthesis of a series of thiosemicarbazones.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Justin W Hicks et al.
Chemistry & biodiversity, 5(11), 2415-2422 (2008-11-28)
Addition of thiosemicarbazide, 4-allylthiosemicarbazide, and 4-phenylthiosemicarbazide to (formylphenyl)boronic acids affords a series of thiosemicarbazones containing boronic acids. Addition of 2-formylphenylboronic acid to the thiosemicarbazides gave the corresponding cyclic 2,3,1-benzodiazaborines. All new compounds have been investigated for potential antifungal activity.
Derya Kara et al.
Journal of hazardous materials, 165(1-3), 1165-1169 (2008-12-17)
A matrix separation and analyte preconcentration system using Amberlite XAD copolymer resins functionalized by Schiff base reactions coupled with atomic spectrometry has been developed. Three different functionalized Amberlite XAD resins were synthesized using 4-phenylthiosemicarbazide, 2,3-dihydroxybenzaldehyde and 2-thiophenecarboxaldehyde as reagents. These

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service