Skip to Content
Merck
Magnesium foil, thickness 0.075 mm, 4 mm diameter, purity 99.9%

Magnesium

Magnesium foil, thickness 0.075 mm, 4 mm diameter, purity 99.9%
Synonym(s):
Magnesium
Empirical Formula (Hill Notation):
Mg
CAS Number:
Molecular Weight:
24.31

Products

Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Diameter (mm)
Length (mm)
Element
Quality Segment
Showing 1-20 of 201
Page 1 of 11
Compare
Product Number
Description
Pricing
Page 1 of 11