Skip to Content
Merck
All Photos(1)

Key Documents

128392

Sigma-Aldrich

1-Fluoro-3-nitrobenzene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4NO2
CAS Number:
Molecular Weight:
141.10
Beilstein:
1862210
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.525 (lit.)

bp

205 °C (lit.)

mp

1.7 °C (lit.)

density

1.325 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

[O-][N+](=O)c1cccc(F)c1

InChI

1S/C6H4FNO2/c7-5-2-1-3-6(4-5)8(9)10/h1-4H

InChI key

WMASLRCNNKMRFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Fluoro-3-nitrobenzene was used as an internal standard in the regiospecific silver-mediated fluorination of aryl silanes.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

168.8 °F

Flash Point(C)

76 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pingping Tang et al.
Tetrahedron, 67(24), 4449-4454 (2011-06-22)
A regiospecific silver-mediated fluorination of aryl silanes is reported. The reaction is operationally simple, and employs Ag(2)O as readily available, inexpensive silver source, which can be recovered.
Bartoli G, et al.
Journal of the Chemical Society. Perkin Transactions 1, 2671-2671 (1972)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service