Skip to Content
Merck
All Photos(1)

Key Documents

156248

Sigma-Aldrich

Sodium ethoxide

95%

Synonym(s):

Sodium ethylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2ONa
CAS Number:
Molecular Weight:
68.05
Beilstein:
3593646
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor density

1.6 (vs air)

Quality Level

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

95%

form

powder

SMILES string

[Na+].CC[O-]

InChI

1S/C2H5O.Na/c1-2-3;/h2H2,1H3;/q-1;+1

InChI key

QDRKDTQENPPHOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium ethoxide (Sodium ethylate) is a sodium alkoxide. It has been synthesized by reacting sodium with ethanol. It undergoes decomposition in the presence of water to afford ethanol and sodium hydroxide. It is widely employed as a strong base in organic synthesis studies.

Application


  • Organic solvent fractionation in lignin depolymerization: A study focused on enhancing the monophenolic yield of lignin depolymerization using a dualistic aprotic solvent system, employing sodium ethoxide for effective organic solvent fractionation. This approach underscores the versatility of sodium ethoxide in complex organic synthesis and biomass processing (Xu et al., 2024).

  • Solid-state self-quenching-resistant sulfur dots: Research on sulfur dots utilized a solvent-type passivation strategy to control solid-state self-quenching-resistant behavior, where sodium ethoxide played a crucial role in the synthesis process. This study illustrates sodium ethoxides utility in advanced materials synthesis, particularly for optoelectronic applications (Wu et al., 2022).

  • Synthesis of corrosion inhibitors: Sodium ethoxide was used in the efficient synthesis of 6,7-Dihydro-5H-cyclopenta[b]pyridine-3-carbonitrile compounds, demonstrating its applicability as a base in the synthesis of corrosion inhibitors for steel alloys. This application combines computational and practical approaches to highlight sodium ethoxide′s role in materials science and corrosion prevention (Abd El-Lateef et al., 2022).

  • Colon-targeted drug delivery systems: The design and synthesis of Thieno[2,3-b]pyridines-chitosan nanocomposites for colon targeting employed sodium ethoxide. This research showcases sodium ethoxides application in the preparation of targeted drug delivery systems, focusing on enhancing the efficacy and specificity of pharmaceutical treatments (Mansour et al., 2020).

  • Synthesis of antimicrobial compounds: Sodium ethoxide was integral in the synthesis of novel bis-alpha,beta-unsaturated ketones and nicotinonitrile derivatives, aimed at antimicrobial applications. This study highlights the compound′s critical role in the development of new antimicrobial agents, demonstrating its utility in medicinal chemistry (Altalbawy, 2013).

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Self-heat. 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(F)

86.0 °F - closed cup

Flash Point(C)

30 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James E Clark et al.
Circulation research, 93(2), e2-e8 (2003-07-05)
Carbon monoxide, which is generated in mammals during the degradation of heme by the enzyme heme oxygenase, is an important signaling mediator. Transition metal carbonyls have been recently shown to function as carbon monoxide-releasing molecules (CO-RMs) and to elicit distinct
Charanjit S Sethi et al.
Investigative ophthalmology & visual science, 46(1), 329-342 (2004-12-30)
To investigate glial remodeling and neuronal plasticity in adult human retinal detachment complicated by proliferative vitreoretinopathy (PVR) and to grade pathologic changes with a severity scoring system. Sixteen full-thickness retinectomy specimens obtained at retinal relaxing surgery for PVR were fixed
Eagleson M.
Concise Encyclopedia Chemistry, 997-997 (1994)
S H Brorson
Micron (Oxford, England : 1993), 32(2), 101-105 (2000-08-11)
The study's purpose was to obtain improved "deplasticizing" of epoxy sections for immunoelectron microscopy. Epoxy-embedded renal swine tissue with immune complex deposits was used. Ultrathin sections were mounted on uncoated grids or on carbon-stabilized formvar grids. The sections were exposed
Francesca Monticelli et al.
Journal of endodontics, 32(5), 447-451 (2006-04-25)
The influence of different silane/dentin adhesives on the microtensile bond strengths of a hybrid composite to sodium ethoxide-etched quartz fiber posts was investigated. A prehydrolyzed silane was applied without an adhesive and compared to two-component systems in which hydrolysis of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service