Skip to Content
Merck
All Photos(2)

Key Documents

128341

Sigma-Aldrich

Fluoroacetamide

Synonym(s):

NSC 31876

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FCH2CONH2
CAS Number:
Molecular Weight:
77.06
Beilstein:
1739054
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

mp

106-108 °C (lit.)

functional group

amide
fluoro

SMILES string

NC(=O)CF

InChI

1S/C2H4FNO/c3-1-2(4)5/h1H2,(H2,4,5)

InChI key

FVTWJXMFYOXOKK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fluoroacetamide is a rodenticide that can be assayed by gas chromatography/mass spectrometry.

Application

Reactant involved in:
  • C-N coupling to amides and nitrogen-containing heterocycles
  • Substitution in aryl triflates

Used in mass databases for screening of pesticide residues via fast liquid chromatography TOF mass spectrometry

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaowei Zhu et al.
International journal of molecular sciences, 20(11) (2019-06-13)
Cuticular wax is a mixture of very long chain fatty acids (VLCFAs) and their derivatives, which determines vital roles for plant growth. In cabbage, the cuticular wax content of leaf blades is an important trait influencing morphological features of the
Sergey V Kuznetsov et al.
Journal of applied toxicology : JAT, 27(6), 538-550 (2007-03-14)
A study was conducted of acute intoxication of infant and adult Wistar rats with fluoroacetamide (FAA), an inhibitor of oxidative metabolism. FAA was administered orally to adult rats at 1/2 LD(50) and subcutaneously to infant rats at LD(100) or 1/10
U Taitelman et al.
Archives of toxicology. Supplement. = Archiv fur Toxikologie. Supplement, 6, 228-231 (1983-01-01)
Two cases are reported of severe acute fluoroacetamide poisoning in man, with successful treatment of the life-threatening cardiac arrhythmias by the administration of calcium chloride. The arrhythmias were preceded by prolongation of the QT interval in the ECG. Calcium chloride
P J Kostyniak et al.
Toxicology letters, 22(2), 217-222 (1984-08-01)
2,4-Dinitrofluorobenzene (DNFB) reacts with glutathione to form a stable product similar to that formed with the model glutathione-S-transferase (GST) substrate, 1-chloro-2,4-dinitrobenzene (CDNB). DNFB is approx. 40 times als reactive as CDNB in this chemical reaction. The enzymatic defluorination of DNFB
Arun K Ghosh et al.
The Journal of organic chemistry, 74(10), 3689-3697 (2009-04-14)
Synthesis and reactivity of N-methoxy-N-methyl-(1,3-benzothiazol-2-ylsulfonyl)fluoroacetamide, a building block for Julia olefination, is reported. This reagent undergoes condensation reactions with aldehydes and cyclic ketones to give alpha-fluorovinyl Weinreb amides. Olefination reactions proceed under mild, DBU-mediated conditions, or in the presence of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service