Skip to Content
Merck
All Photos(3)

Key Documents

126179

Sigma-Aldrich

9,10-Dihydroanthracene

97%

Synonym(s):

Dihydroanthracene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12
CAS Number:
Molecular Weight:
180.25
Beilstein:
1364575
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

bp

312 °C (lit.)

mp

103-107 °C (lit.)

density

0.88 g/mL at 25 °C (lit.)

SMILES string

C1c2ccccc2Cc3ccccc13

InChI

1S/C14H12/c1-2-6-12-10-14-8-4-3-7-13(14)9-11(12)5-1/h1-8H,9-10H2

InChI key

WPDAVTSOEQEGMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

9,10-Dihydroanthracene causes the transfer hydrogenation of C60 and C70 in the presence of [7H]benzanthrene catalyst. It was oxidatively aromatized to the corresponding anthracene in the presence of molecular oxygen as an oxidant and activated carbon as a promoter in xylene.

Application

9,10-Dihydroanthracene(DHA) has been used in a study to assess the hydrogen abstraction capability of valence-delocalized iron complex with DHA in MeCN.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nirupama Singh et al.
Journal of inorganic biochemistry, 197, 110674-110674 (2019-05-21)
Developing coordination complexes of earth abundant metals that can perform substrate oxidations under benign conditions is an ongoing challenge. Herein, the reactivity of two mononuclear Cu-complexes toward the oxidant H2O2 is reported. Both complexes displayed ligand oxidation upon reaction with
Genqiang Xue et al.
Proceedings of the National Academy of Sciences of the United States of America, 104(52), 20713-20718 (2007-12-21)
Intermediate Q, the methane-oxidizing species of soluble methane monooxygenase, is proposed to have an [Fe(IV)(2)(mu-O)(2)] diamond core. In an effort to obtain a synthetic precedent for such a core, bulk electrolysis at 900 mV (versus Fc(+/0)) has been performed in
[7H] Benzanthrone, a catalyst for the transfer hydrogenation of C60 and C70 by 9, 10-dihydroanthracene.
Gerst M, et al.
Tetrahedron Letters, 34(48), 7729-7732 (1993)
Pengfei Sun et al.
Small (Weinheim an der Bergstrasse, Germany), 16(3), e1906462-e1906462 (2019-12-24)
Lithium primary batteries are still widely used in military, aerospace, medical, and civilian applications despite the omnipresence of rechargeable Li-ion batteries. However, these current primary chemistries are exclusively based on inorganic materials with high cost, low energy density or severe
Natsuki Nakamichi et al.
The Journal of organic chemistry, 68(21), 8272-8273 (2003-10-11)
Substituted 9,10-dihydroanthracenes were oxidatively aromatized to the corresponding anthracenes effectively by using molecular oxygen as an oxidant and activated carbon (Darco KB, Aldrich, Inc.) as a promoter in xylene.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service