Skip to Content
Merck
All Photos(1)

Key Documents

123889

Sigma-Aldrich

α,α′-Dichloro-o-xylene

98%

Synonym(s):

1,2-Bis(chloromethyl)benzene, o-Xylylene dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH2Cl)2
CAS Number:
Molecular Weight:
175.06
Beilstein:
2043675
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

239-241 °C (lit.)

mp

51-55 °C (lit.)

functional group

chloro

SMILES string

ClCc1ccccc1CCl

InChI

1S/C8H8Cl2/c9-5-7-3-1-2-4-8(7)6-10/h1-4H,5-6H2

InChI key

FMGGHNGKHRCJLL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α,α′-Dichloro-o-xylene reacts with elemental tellurium and NaI in 2-methoxyethanol to form 1,1-diiodo-3,4-benzo-1-telluracyclopentane.

Application

α,α′−Dichloro-o-xylene was used in solid-phase synthesis of large combinatorial variations of fundamental peptide unit.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

257.0 °F - closed cup

Flash Point(C)

125 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The synthesis and characterization of α-and β-1, 1-diiodo-3, 4-benzo-1-telluracyclopentane, C8 H8Tel2.
Ziolo RF and Gunther WHH.
Journal of Organometallic Chemistry, 146(3), 245-251 (1978)
William L Scott et al.
Molecules (Basel, Switzerland), 15(7), 4961-4983 (2010-07-27)
Amino acids are Nature's combinatorial building blocks. When substituted on both the amino and carboxyl sides they become the basic scaffold present in all peptides and proteins. We report a solid-phase synthetic route to large combinatorial variations of this fundamental

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service