Skip to Content
Merck
All Photos(2)

Key Documents

113182

Sigma-Aldrich

1-Phenylheptane

98%

Synonym(s):

Heptylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5(CH2)6CH3
CAS Number:
Molecular Weight:
176.30
Beilstein:
1905782
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.485 (lit.)

bp

233 °C (lit.)

mp

−48 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

functional group

phenyl

SMILES string

CCCCCCCc1ccccc1

InChI

1S/C13H20/c1-2-3-4-5-7-10-13-11-8-6-9-12-13/h6,8-9,11-12H,2-5,7,10H2,1H3

InChI key

LBNXAWYDQUGHGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Heptylbenzene (1-Phenylheptane) forms charge-transfer complexes with fluoranil and 1,3,5-trinitrobenzene.

Application

1-Phenylheptane is used in zirconia and titania based stationary phase capillary HTLC, to separate alkyl benzene derivatives using an acetonitrile-water eluent.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giorgia Purcaro et al.
Analytica chimica acta, 1027, 158-167 (2018-06-06)
Untargeted metabolomics study of volatile organic compounds produced by different cell cultures is a field that has gained increasing attention over the years. Solid-phase microextraction has been the sampling technique of choice for most of the applications mainly due to
Hot eluent capillary liquid chromatography using zirconia and titania based stationary phases.
Kephart TS, et al.
Analytica Chimica Acta, 414(1-2), 71-78 (2000)
K Elias Ikonen et al.
Journal of chromatography. A, 1614, 460705-460705 (2019-11-23)
An update for ASTM D2425, "Standard Test Method for Hydrocarbon Types in Middle Distillates by Mass Spectrometry," was recently released to expand its scope by adding modern mass spectrometric instrumentation and a new sample type, synthesized hydrocarbons. While the method

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service