Skip to Content
Merck
All Photos(1)

Key Documents

113158

Sigma-Aldrich

4-Methoxy-α-toluenethiol

90%, technical grade

Synonym(s):

4-Methoxybenzyl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH2SH
CAS Number:
Molecular Weight:
154.23
Beilstein:
471686
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

refractive index

n20/D 1.573 (lit.)

bp

90-95 °C/0.5 mmHg (lit.)

density

1.107 g/mL at 25 °C (lit.)

functional group

thiol

storage temp.

2-8°C

SMILES string

COc1ccc(CS)cc1

InChI

1S/C8H10OS/c1-9-8-4-2-7(6-10)3-5-8/h2-5,10H,6H2,1H3

InChI key

PTDVPWWJRCOIIO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Methoxy-α-toluenethiol is used as an internal standard in the Quantitative determination of wine polyfunctional mercaptans at the level of nanogram per liter.
4-methoxy-α-toluenethiol is used to synthesize many ω-mercapto amino acids with side-chain lengths ranging from 3–5 methylene units.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of protected ω-mercapto amino acids: precursors for incorporation of elongated cysteines into peptides.
Tetrahedron Asymmetry, 9(15), 2739-2743 (1998)
Laura Mateo-Vivaracho et al.
Journal of chromatography. A, 1146(2), 242-250 (2007-02-24)
A fast method for the determination of aroma-powerful polyfunctional thiols at nanogram per liter level has been developed and applied to wine. A small volume of wine (6 mL) was extracted with 1.5 mL of benzene containing four internal standards.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service