Skip to Content
Merck
All Photos(4)

Key Documents

T28525

Sigma-Aldrich

4-Methylbenzenethiol

98%

Synonym(s):

p-Thiocresol, 4-Mercaptotoluene, 4-Methylthiophenol, p-Toluenethiol, p-Tolyl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SH
CAS Number:
Molecular Weight:
124.20
Beilstein:
605761
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

6.2 (vs air)

Quality Level

vapor pressure

10 mmHg ( 71 °C)

Assay

98%

bp

195 °C (lit.)

mp

41-43 °C (lit.)

SMILES string

Cc1ccc(S)cc1

InChI

1S/C7H8S/c1-6-2-4-7(8)5-3-6/h2-5,8H,1H3

InChI key

WLHCBQAPPJAULW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Han Lu et al.
Nanoscale research letters, 15(1), 8-8 (2020-01-15)
Fabrication of large-area devices with patternable nanostructures is important for practical applications in optical or electrical devices. In this work, we describe an easy and environment-friendly method for preparing large-area nano-dot (ND) arrays via the electrolytic reaction of a metal
Nicolás Arisnabarreta et al.
Langmuir : the ACS journal of surfaces and colloids, 33(48), 13733-13739 (2017-11-08)
The effect of the Au crystalline plane on the adsorption of different thiols and selenols is studied via reductive desorption (RD) and X-ray photoelectron spectroscopy (XPS) measurements. Self-assembled monolayers (SAMs) using aliphatic (ATs) and aromatic thiols (ArTs) on both Au(111)
Camilla I Nantes et al.
ChemMedChem, 15(5), 449-458 (2019-12-14)
Organosulfur compounds show cytotoxic potential towards many tumor cell lines. Disulfides and thiosulfonates act through apoptotic processes, inducing proteins associated with apoptosis, endoplasmic reticulum stress, and the unfolded protein response. Three p-substituted symmetric diaryl disulfides and three diaryl thiosulfonates were
Chatdanai Lumdee et al.
Nanoscale, 7(9), 4250-4255 (2015-02-13)
The effect of nanoscale surface roughness on the gap plasmon resonance of gold nanoparticles on thermally evaporated gold films is investigated experimentally and numerically. Single-particle scattering spectra obtained from 80 nm diameter gold particles on a gold film show significant
Suhailah S Al-Jameel et al.
World journal of microbiology & biotechnology, 34(4), 52-52 (2018-03-20)
Hay synthesis of a novel series of symmetrically tetra-substituted thiophenyl zinc(II)phthalocyanines (RS)4ZnPcs 4a-c was reported. Their novel 4-thiophenyl-phthalonitriles precursors 3(a-c) were synthesized from their substituted thiophenols 2(a-c). They were screened for their in-vitro antitumor activity on Human lung adenocarcinoma (A549)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service