Skip to Content
Merck
All Photos(2)

Key Documents

112917

Sigma-Aldrich

2-Methyl-1-propanethiol

92%, technical grade

Synonym(s):

Isobutyl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2SH
CAS Number:
Molecular Weight:
90.19
Beilstein:
1730890
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

vapor density

3.1 (vs air)

vapor pressure

124 mmHg ( 37.8 °C)

Assay

92%

refractive index

n20/D 1.4385 (lit.)

bp

87-89 °C (lit.)

solubility

H2O: slightly soluble
alcohol: very soluble
diethyl ether: very soluble

density

0.831 g/mL at 25 °C (lit.)

functional group

thiol

SMILES string

CC(C)CS

InChI

1S/C4H10S/c1-4(2)3-5/h4-5H,3H2,1-2H3

InChI key

BDFAOUQQXJIZDG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-methyl-1-propanethiol attaches with L-Cys (the carrier) via a disulfide bond to form the brain-targeted drug delivery systems (BTDS).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

14.0 °F - closed cup

Flash Point(C)

-10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dennis M Killian et al.
Drug delivery, 14(1), 25-31 (2006-11-17)
We describe a novel strategy to achieve high affinity recognition for the specific, cerebrovascular large neutral amino acid transporter (LAT1) isoform by covalent coupling of small molecules to the amino acid, L-cysteine (L-Cys). L-Cys (as the carrier) was covalently attached

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service