Merck
  • Medial plantar nerve ligation as a novel model of neuropathic pain in mice: pharmacological and molecular characterization.
Medial plantar nerve ligation as a novel model of neuropathic pain in mice: pharmacological and molecular characterization.
Scientific reports (2016-05-28)
Morena B Sant'Anna, Ricardo Kusuda, Tiago A Bozzo, Gabriel S Bassi, José C Alves-Filho, Fernando Q Cunha, Sergio H Ferreira, Guilherme R Souza, Thiago M Cunha
ABSTRACT
Peripheral neuropathic pain is a consequence of an injury/disease of the peripheral nerves. The mechanisms involved in its pathophysiology are not entirely understood. To better understand the mechanisms involved in the development of peripheral nerve injury-induced neuropathic pain, more experimental models are required. Here, we developed a novel peripheral neuropathic pain model in mice by using a minimally invasive surgery and medial plantar nerve ligation (MPNL). After MPNL, mechanical allodynia was established, and mice quickly recovered from the surgery without any significant motor impairment. MPNL causes an increased expression of ATF-3 in the sensory neurons. At 14 days after surgery, gabapentin was capable of reversing the mechanical allodynia, whereas anti-inflammatory drugs and opioids were ineffective. MPNL-induced neuropathic pain was mediated by glial cells activation and the production of TNF-α and IL-6 in the spinal cord. These results indicate MPNL as a reasonable animal model for the study of peripheral neuropathic pain, presenting analgesic pharmacological predictivity to clinically used drugs. The results also showed molecular phenotypic changes similar to other peripheral neuropathic pain models, with the advantage of a lack of motor impairment. These features indicate that MPNL might be more appropriate for the study of neuropathic pain than classical models.
Sigma-Aldrich
Dexamethasone 21-phosphate disodium salt, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, Chemicon®, from chicken
Pricing and availability is not currently available.
Roche
cOmplete Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Pricing and availability is not currently available.