Skip to Content
Merck
All Photos(2)

Key Documents

04693116001

Roche

cOmplete Protease Inhibitor Cocktail

Tablets provided in EASYpacks

Synonym(s):

protease inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.21
form:
tablet
solubility:
water: soluble
storage temp.:
2-8°C

form

tablet

packaging

pkg of 20 tablets

manufacturer/tradename

Roche

solubility

water: soluble

absorption

0.08 at 280 nm

storage temp.

2-8°C

General description

cOmplete Protease Inhibitor Tablets inhibit a broad spectrum of serine, cysteine, and metalloproteases, as well as calpains. Due to the optimized composition of the tablets, they show excellent inhibition effects, and are well suited for the protection of proteins isolated from animal tissues, plants, yeast, and bacteria.
cOmplete tablets contain EDTA and both irreversible and reversible protease inhibitors. Protease inhibitors present in cOmplete Tablets do not form irreversible complexes with the SH groups of proteins.

Specificity

Inhibits a broad spectrum of serine proteases, cysteine proteases, and metalloproteases, as well as calpains.

Application

cOmplete Protease Inhibitor Cocktail has been used as an component in
  • extraction buffer for protein extraction from Arabidopsis seedling
  • in radioimmunoprecipitation assay (RIPA) buffer for the isolation of protein from lymphocytes
  • the extraction buffer for the lysis of protoplast.

cOmplete is used for the inhibition of serine, cysteine, and metalloproteases in bacterial, mammalian, yeast, and plant cell extracts.
cOmplete contains both reversible and irreversible protease inhibitors. Therefore, we recommend the addition of cOmplete to all stock buffers and solutions normally protected with protease inhibitors, and not  just during the initial purification steps.
cOmplete contains EDTA (1 tablet yields a  1 mM EDTA solution  in 50 ml). Therefore, the extraction buffer should not contain divalent cations like Ca2+, Mg2+, or Mn2+, otherwise the inhibition of the metalloproteases might be incomplete. If the protein of interest will be purified by IMAC (Immobilized Metal Affinity Chromatography), for example, Poly-His-tagged recombinant proteins, EDTA has to be eliminated (e.g., by dialysis) prior to the chromatography. Alternatively, the product cOmplete, EDTA-free, can be used. These tablets are identical to cOmplete, with the exception that no EDTA or other chelating agent is  present.
cOmplete Tablets inhibit proteolytic activity in large extraction volumes (up to 50 ml). If you are working with smaller volumes (up to 10 ml), we recommend to use cOmplete, Mini. The composition of the cOmplete, Mini Tablet, is identical to the normal cOmplete Tablet.

Features and Benefits

Use cOmplete Protease Inhibitor Tablets to protect your proteins from a wide range of proteases. In just minutes, uninhibited proteolytic activity can degrade the protein you have spent days isolating. Each of these convenient water-soluble tablets contains a blend of protease inhibitors that inhibits proteolytic activity from most cell types, including animals, plants, yeast, and bacteria. No weighing or measuring is necessary, ensuring consistent results.

  • Convenience: Simply push the quick-dissolving tablet through the foil packaging of the EASYpack into your buffer.
  • Complete protection: Instantly protect your proteins against a broad range of proteases.
  • Flexible: Protect proteins in extracts from almost any tissue or cell, including animals, plants, yeast, bacteria, or fungi.
  • Safe: Choose non-toxic inhibitors that pose no risk to you or those around you.

Packaging

20 individually packed cOmplete Protease Inhibitor Cocktail Tablets in foil blisters.

Components

Proprietary mixture of several protease inhibitors with broad inhibitory specificity.

Physical form

Tablets in foil blister packs

Preparation Note

Working concentration: 1 tablet per 50ml extraction solution
Working solution: Handling

Carefully push the tablet through the foil packaging using the base of your thumb (not finger nail) to prevent the breakage of tablets.

Preparation of Working Solution

One cOmplete tablet is sufficient for the inhibition of the proteolytic activity in 50ml extraction solution. When a very high proteolytic activity is present, one tablet should be used for 25ml extraction buffer. Tablets can be added directly to the extraction medium. Alternatively, a stock solution (25 X conc.) can be prepared.

Preparation of Stock solution (25 X conc.)

Dissolve one cOmplete tablet in 2ml dist. water or in 2ml 100 mM phosphate buffer, pH 7.0.
Storage conditions (working solution): The stock solution can be stored at 2 to 8°C for 1 to 2 weeks, or at least 12 weeks at -15 to -25°C.

Reconstitution

Soluble in aqueous buffers, or add directly to extraction media. Alternatively, prepare 25x stock solutions in 2 ml water or 100 mM phosphate buffer, pH 7.0. Stock solution is stable for 1-2 weeks at 2 to 8 °C or at least 12 weeks at -15 to -25 °C; can be used in thiol-containing solutions at 15 to 25 °C.

Storage and Stability

Store at 2 to 8 °C. (storage at -15 to -25 °C is also possible)

Legal Information

cOmplete is a trademark of Roche

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanke Chen et al.
Nature cell biology, 22(10), 1276-1285 (2020-10-03)
Breast cancer brain metastasis (BCBM) is a devastating disease. Radiation therapy remains the mainstay for treatment of this disease. Unfortunately, its efficacy is limited by the dose that can be safely applied. One promising approach to overcoming this limitation is
Mingwei Ni et al.
Glia, 59(5), 810-820 (2011-02-26)
As the two major glial cell types in the brain, astrocytes and microglia play pivotal but different roles in maintaining optimal brain function. Although both cell types have been implicated as major targets of methylmercury (MeHg), their sensitivities and adaptive
EL1-like casein kinases suppress ABA signaling and responses by phosphorylating and destabilizing the ABA receptors PYR/PYLs in Arabidopsis
Chen HH, et al.
Molecular plant, 11(5), 706-719 (2018)
The receptor-like cytoplasmic kinase BIK1 localizes to the nucleus and regulates defense hormone expression during plant innate immunity
Lal NK, et al.
Cell host & microbe, 23(4), 485-497 (2018)
Sinead Toomey et al.
Journal of translational medicine, 18(1), 99-99 (2020-02-24)
An increasing number of anti-cancer therapeutic agents target specific mutant proteins that are expressed by many different tumor types. Successful use of these therapies is dependent on the presence or absence of somatic mutations within the patient's tumor that can

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service