Skip to Content
Merck
All Photos(5)

Key Documents

I7378

Sigma-Aldrich

Indomethacin

98.5-100.5% (in accordance with EP), powder or crystals, COX inhibitor

Synonym(s):

1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-3-indoleacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H16ClNO4
CAS Number:
Molecular Weight:
357.79
Beilstein:
497341
EC Number:
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

Product Name

Indomethacin, 98.5-100.5% (in accordance with EP)

biological source

synthetic

Quality Level

Assay

98.5-100.5% (in accordance with EP)

form

powder or crystals

loss

≤0.5% loss on drying

mp

158-162 °C

Mode of action

enzyme | inhibits

SMILES string

COc1ccc2n(c(C)c(CC(O)=O)c2c1)C(=O)c3ccc(Cl)cc3

InChI

1S/C19H16ClNO4/c1-11-15(10-18(22)23)16-9-14(25-2)7-8-17(16)21(11)19(24)12-3-5-13(20)6-4-12/h3-9H,10H2,1-2H3,(H,22,23)

InChI key

CGIGDMFJXJATDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Indomethacin is an inhibitor of the enzyme cyclooxygenase 1 and 2. Indomethacin elicits side effects in gastrointestinal tract, kidney and cerebrum. Indomethacin, along with ibuprofen, is effective for treating patent ductus arteriosus (PDA) in infants with respiratory distress syndrome. Rectal indomethacin is effective in treating endoscopic retrograde cholangiography (ERCP) induced pancreatitis.

Application

Indomethacin has been used:
  • as a medium supplement in osteogenic and adipogenic differentiation assays in human bone marrow stem cells
  • as a medium supplement in bovine amniotic fluid stem cells (BAFSCs) culture
  • in the inhibition of prostaglandin E2 (PGE2) in T cells

Biochem/physiol Actions

Cyclooxygenase (COX) inhibitor that is relatively selective for COX-1.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Prostanoid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A comparison of ibuprofen and indomethacin for closure of patent ductus arteriosus
Van Overmeire B, et al.
The New England Journal of Medicine, 343(10), 674-681 (2000)
Therapeutic potential of amniotic fluid stem cells to treat bilateral ovarian dystrophy in dairy cows in a subtropical region
Chang LB, et al.
Reproduction in domestic animals = Zuchthygiene, 53(2), 433-441 (2018)
Suppression of T cells by mesenchymal and cardiac progenitor cells is partly mediated via extracellular vesicles
van den Akker F, et al.
Heliyon, 4(6), e00642-e00642 (2018)
Effectiveness of the use of Rectal Indometacin in the prevention of post-CPRE pancreatitis in the Naval Medical Center.
Islava EE, et al.
INTERNATIONAL JOURNAL OF ADVANCED MULTIDISCIPLINARY RESEARCH, 5(11), 44-51 (2018)
Modulation of miRNAs by Vitamin C in Human Bone Marrow Stromal Cells
Kolhe R, et al.
Nutrients, 10(2), 186-186 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service