Merck
Heterolysis of Dihydrogen by Silver Alkoxides and Fluorides.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-06-11)
Brandon K Tate, Jenna T Nguyen, John Bacsa, Joseph P Sadighi
ABSTRACT
Alkoxide-bridged disilver cations react with dihydrogen to form hydride-bridged cations, releasing free alcohol. Hydrogenolysis of neutral silver fluorides affords hydride-bridged disilver cations as their bifluoride salts. These reactions proceed most efficiently when the supporting ligands are expanded N-heterocyclic carbenes (NHCs) derived from 6- and 7-membered cyclic amidinium salts. Kinetics studies show that silver fluoride hydrogenolysis is first-order in both silver and dihydrogen.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, contains no stabilizer
Sigma-Aldrich
Tetrahydrofuran, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium tert-pentoxide solution, 1.4 M in THF
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Lithium aluminum hydride, hydrogen-storage grade
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Sodium tert-pentoxide solution, 40% in toluene
Sigma-Aldrich
Rhein
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
1,4-Dimethoxybenzene, ReagentPlus®, 99%
Sigma-Aldrich
Lithium aluminum hydride, ≥97.0% (gas-volumetric)
Sigma-Aldrich
Lithium aluminum hydride, powder, reagent grade, 95%
Sigma-Aldrich
1,4-Dimethoxybenzene, 99%, FG
Sigma-Aldrich
Lithium aluminum hydride, pellets, reagent grade, 95%
Sigma-Aldrich
Rhein, technical grade
Sigma-Aldrich
Argon, ≥99.998%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, SAJ first grade, ≥99.0%
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Sigma-Aldrich
Calcium hydride, purum p.a., ≥97.0% (gas-volumetric), powder
Sigma-Aldrich
Calcium hydride, powder, 0-2 mm, reagent grade, ≥90% (gas-volumetric)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Sodium tert-pentoxide, 95%
Sigma-Aldrich
Pentane, anhydrous, ≥99%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent