Skip to Content
Merck
All Photos(2)

Key Documents

343773

Sigma-Aldrich

Deuterium oxide

99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)

Synonym(s):

Heavy water, Water-d2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
D2O
CAS Number:
Molecular Weight:
20.03
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21
technique(s):
NMR: suitable
bp:
101.4 °C (lit.)

isotopic purity

99.9 atom % D

Quality Level

form

liquid

contains

1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)

technique(s)

NMR: suitable

bp

101.4 °C (lit.)

mp

3.8 °C (lit.)

SMILES string

[2H]O[2H]

InChI

1S/H2O/h1H2/i/hD2

InChI key

XLYOFNOQVPJJNP-ZSJDYOACSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Deuterium oxide can serve as a solvent in the :
  • NMR analysis.
  • 1HDOSY (Diffusion-Ordered Spectroscopy) NMR experiments.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Structure of water and hydrophobic bonding in proteins. IV. The thermodynamic properties of liquid deuterium oxide.
Nemethy G and Scheraga HA.
J. Chem. Phys. , 41(3), 680-689 (1964)
The ionization constant of deuterium oxide from 5 to 50?.
Covington AK, et al.
The Journal of Physical Chemistry, 70(12), 3820-3824 (1966)
Using high-performance quantitative NMR (HP-qNMR?) for certifying traceable and highly accurate purity values of organic reference materials with uncertainties< 0.1%.
Weber M, et al.
Accreditation and Quality Assurance, 18(2), 91-98 (2013)
Liset Westera et al.
Blood, 122(13), 2205-2212 (2013-08-16)
Quantitative knowledge of the turnover of different leukocyte populations is a key to our understanding of immune function in health and disease. Much progress has been made thanks to the introduction of stable isotope labeling, the state-of-the-art technique for in
Lars Holm et al.
American journal of physiology. Endocrinology and metabolism, 304(8), E895-E907 (2013-02-21)
A method to determine the rate of protein breakdown in individual proteins was developed and tested in rats and confirmed in humans, using administration of deuterium oxide and incorporation of the deuterium into alanine that was subsequently incorporated into body

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service