Merck
  • Regulation of homologous recombinational repair by lamin B1 in radiation-induced DNA damage.
Regulation of homologous recombinational repair by lamin B1 in radiation-induced DNA damage.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2015-03-04)
Ning-Ang Liu, Jiying Sun, Kazuteru Kono, Yasunori Horikoshi, Tsuyoshi Ikura, Xing Tong, Tokuko Haraguchi, Satoshi Tashiro
ABSTRACT
DNA double-strand breaks (DSBs) are the major lethal lesion induced by ionizing radiation (IR). RAD51-dependent homologous recombination (HR) is one of the most important pathways in DSB repair and genome integrity maintenance. However, the mechanism of HR regulation by RAD51 remains unclear. To understand the mechanism of RAD51-dependent HR, we searched for interacting partners of RAD51 by a proteomics analysis and identified lamin B1 in human cells. Lamins are nuclear lamina proteins that play important roles in the structural organization of the nucleus and the regulation of chromosome functions. Immunoblotting analyses revealed that siRNA-mediated lamin B1 depletion repressed the DNA damage-dependent increase of RAD51 after IR. The repression was abolished by the proteasome inhibitor MG132, suggesting that lamin B1 stabilizes RAD51 by preventing proteasome-mediated degradation in cells with IR-induced DNA damage. We also showed that lamin B1 depletion repressed RAD51 focus formation and decreased the survival rates after IR. On the basis of these results, we propose that lamin B1 promotes DSB repair and cell survival by maintaining the RAD51 protein levels for HR upon DSB induction after IR.
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Anti-Lamin A−Atto 647N antibody produced in rabbit, 1.5-3 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
GeneJuice® Transfection Reagent, Non-lipid based chemical transfection reagent optimized for maximum transfection efficiency, ease-of-use, and minimal cytotoxicity on a wide variety of mammalian cells.
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard