Merck
  • Improved sensitivity gas chromatography-mass spectrometry determination of parabens in waters using ionic liquids.
Improved sensitivity gas chromatography-mass spectrometry determination of parabens in waters using ionic liquids.
Talanta (2015-12-24)
Juan Ignacio Cacho, Natalia Campillo, Pilar Viñas, Manuel Hernández-Córdoba
ABSTRACT
A new procedure for the introduction of ionic liquid samples in gas chromatography (GC) is proposed. This procedure, based on microvial insert thermal desorption, allows the direct analysis of the compounds preconcentrated by ionic liquid based liquid-liquid microextraction (IL-LLME) using the combination of a thermal desorption unit (TDU) and a programmed temperature vaporization injector (PTV). Two different IL-LLME methodologies, one based on the formation of a microdroplet emulsion by dispersive liquid-liquid microextraction (DLLME) and other through ultrasound-assisted emulsification microextraction (USAEME) were studied and evaluated. IL-DLLME proved advantageous and consequently, it was adopted for preconcentration purposes. This easy to apply approach was used for the determination of five parabens (methyl-, ethyl-, propyl-, butyl- and isobutyl paraben) in swimming pool waters, after in situ acetylation. The optimized conditions of TDU/PTV allowed the analytes contained in 20 µL of the enriched IL to be transferred to the capillary column. Quantification of the samples was carried out against aqueous standards, and quantification limits of between 4.3 and 8.1 ng L(-1) were obtained, depending on the compound. Concentrations of between 9 and 47 ng L(-1) for some analytes were obtained in the analysis of ten samples.
Sigma-Aldrich
Isobutyl 4-hydroxybenzoate, 97%
Supelco
Methyl Paraben, PESTANAL®, analytical standard
Sigma-Aldrich
4′-Hydroxyacetophenone, 99%
Sigma-Aldrich
6-Methylpurine, ≥99%
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, with 1% trimethylchlorosilane, for GC derivatization, LiChropur
Sigma-Aldrich
Butyl 4-hydroxybenzoate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Ethyl 4-hydroxybenzoate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetic anhydride, JIS special grade
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ≥99%, FCC
Sigma-Aldrich
Butyl 4-hydroxybenzoate, ≥99.0% (GC)
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, for luminescence, for molecular biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Acetic anhydride, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Butyl 4-hydroxybenzoate, ≥99%
Sigma-Aldrich
N,O-Bis(trimethylsilyl)trifluoroacetamide, ≥99%
Sigma-Aldrich
Ethyl 4-hydroxybenzoate, ReagentPlus®, 99%
Sigma-Aldrich
Acetic anhydride, 99.5%
Sigma-Aldrich
Propyl 4-hydroxybenzoate, ≥99%
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Propyl 4-hydroxybenzoate, BioXtra
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ReagentPlus®, ≥99.0%, crystalline
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Methyl 4-hydroxybenzoate, analytical standard
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Sigma-Aldrich
Potassium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Acetic anhydride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Methyl 4-hydroxybenzoate, SAJ first grade, ≥98.0%
Sigma-Aldrich
Acetic anhydride, ACS reagent, ≥98.0%