09-1970
Ethyl 4-hydroxybenzoate
SAJ first grade, ≥99.0%
Synonym(s):
Ethylparaben
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HOC6H4CO2C2H5
CAS Number:
Molecular Weight:
166.17
Beilstein:
1101972
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
Recommended Products
grade
SAJ first grade
Assay
≥99.0%
availability
available only in Japan
bp
297-298 °C (lit.)
mp
114-117 °C (lit.)
SMILES string
CCOC(=O)c1ccc(O)cc1
InChI
1S/C9H10O3/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6,10H,2H2,1H3
InChI key
NUVBSKCKDOMJSU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Sawsan El Hussein et al.
Experimental dermatology, 16(10), 830-836 (2007-09-12)
Concern is continuously raised about the safety of parabens which are present in most of the cosmetic preparations. In this investigation, methyl-, ethyl-, propyl- and butyl paraben (MP, EP, PP, BP), in a commercial cosmetic lotion, were deposited on human
S Nicoli et al.
Journal of pharmaceutical sciences, 97(11), 4830-4839 (2008-03-05)
This work aims at investigating the nicotinamide (NA)-ethyl-paraben (EP) binary system both in solution and in the solid state. In particular, the apparent EP solubility in water was studied in the presence of different NA concentrations (between 0.28 and 1.64
Shinshi Oishi
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 42(11), 1845-1849 (2004-09-08)
Parabens are alkyl esters of p-hydroxybenzoic acid widely used as preservatives in foodstuffs, cosmetics toiletries and pharmaceuticals. These compounds are known to exert a weak estrogenic activity in estrogen receptor assays in vitro. In addition butyl and propyl parabens show
Suzanne Abbas et al.
Drug metabolism and pharmacokinetics, 25(6), 568-577 (2010-10-12)
Parabens (alkyl esters of 4-hydroxybenzoic acid) are widely used as preservatives in drugs, cosmetic products, and foodstuffs. Safety concerns have recently increased due to the potential health risks associated to exposure to large amounts of these substances. Biotransformation of parabens
Maw-Rong Lee et al.
Journal of chromatography. A, 1120(1-2), 244-251 (2006-03-04)
This study evaluated supercritical fluid extraction (SFE) combined with liquid chromatography-mass spectrometry (LC-MS) to determine trace preservatives and antioxidants including methylparaben (MP), ethylparaben (EP), propylparaben (PP), butylparaben (BP), butylated hydroxyanisole (BHA), butylated hydroxytoluene (BHT), alpha-tocopherol (alpha-t) and alpha-tocopherol acetate (alpha-ta)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service