Skip to Content
Merck
All Photos(3)

Key Documents

P5835

Sigma-Aldrich

Propyl 4-hydroxybenzoate

BioXtra

Synonym(s):

4-Hydroxybenzoic acid propyl ester, Propyl paraben

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CO2CH2CH2CH3
CAS Number:
Molecular Weight:
180.20
Beilstein:
1103245
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

product line

BioXtra

Quality Level

form

powder

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.1%

mp

95-98 °C (lit.)

solubility

ethanol: 0.1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.1%

cation traces

Al: ≤0.0005%
Ca: ≤0.005%
Cu: ≤0.0005%
Fe: ≤0.005%
K: ≤0.01%
Mg: ≤0.001%
NH4+: ≤0.05%
Na: ≤0.01%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

CCCOC(=O)c1ccc(O)cc1

InChI

1S/C10H12O3/c1-2-7-13-10(12)8-3-5-9(11)6-4-8/h3-6,11H,2,7H2,1H3

InChI key

QELSKZZBTMNZEB-UHFFFAOYSA-N

Gene Information

rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

356.0 °F

Flash Point(C)

180 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Herrero et al.
Mutation research, 743(1-2), 20-24 (2012-01-18)
Di(2-ethylhexyl)phthalate, triclosan and propylparaben are contaminants of emerging concern that have been subjected to extensive toxicological studies, but for which limited information is currently available concerning adverse effects on terrestrial plant systems. The Allium cepa test, which is considered one
Michelle P Dvores et al.
Langmuir : the ACS journal of surfaces and colloids, 28(17), 6978-6984 (2012-03-29)
This study presents a method for one-step formation of poly(ethylene oxide) nanofibers incorporating nanoparticles of a poorly water-soluble compound. Using the new method reported here, nanofiber-nanoparticle composites are fabricated in one step by electrospinning of an oil-in-water microemulsion, in which
José Manuel Pérez Martín et al.
Mutation research, 702(1), 86-91 (2010-08-05)
Propyl p-hydroxybenzoate, commonly referred to as propylparaben, is the most frequently used preservative to inhibit microbial growth and extend shelf life of a range of consumer products. The objective of this study was to provide further insight into the toxicological
Dennis G Maki et al.
Critical care medicine, 39(4), 613-620 (2011-01-05)
Catheter-related bloodstream infection is the greatest threat to the safety of patients on hemodialysis. Catheter lock solutions containing heparin have been linked to an increased risk of hemorrhage and thrombocytopenia. To ascertain the safety and efficacy for prevention of catheter-related
C L Barberis et al.
Journal of food protection, 73(8), 1493-1501 (2010-09-08)
Each year, a significant portion of the peanuts produced cannot be marketed because of fungal disease at the postharvest stage and mycotoxin contamination. Antioxidants could be used as an alternative to fungicides to control ochratoxigenic fungi in peanuts during storage.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service