Merck
  • Olfactometry Profiles and Quantitation of Volatile Sulfur Compounds of Swiss Tilsit Cheeses.
Olfactometry Profiles and Quantitation of Volatile Sulfur Compounds of Swiss Tilsit Cheeses.
Journal of agricultural and food chemistry (2015-08-01)
Pascal Fuchsmann, Mireille Tena Stern, Yves-Alain Brügger, Katharina Breme
ABSTRACT
To establish the odor profiles of three differently fabricated commercial Swiss Tilsit cheeses, analyses were conducted using headspace solid-phase microextraction gas chromatography-mass spectrometry/pulsed flame photometric detection and gas chromatography-olfactometry to identify and quantitate volatile compounds. In addition, odor quality and the impact of target sulfur compounds on the overall odor of the cheeses were investigated. The odor profile was found to be mainly influenced by buttery-cheesy and sulfury odor notes in all cheeses. Buttery-cheesy odor notes were attributed to three main molecules: butanoic acid, 3-methylbutanoic acid, and butane-2,3-dione. Over a dozen volatile sulfur compounds were detected at parts per billion levels, but only a few influenced the odor profile of the cheeses: methanethiol, dimethyl disulfide, bis(methylthio)methane, dimethyl trisulfide, 3-(methylthio)propanal, and 2-methyltetrahydrothiophen-3-one (tentative). In conclusion, the conducted analyses allowed differentiation of the cheeses, and gas chromatography-olfactometry results confirmed that partially thermized milk cheese has a more intense and more multifaceted overall flavor.
Product Description
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm(PEG, for use with autosampler
Sigma-Aldrich
Sulfuric acid solution, 0.25 M
Sigma-Aldrich
Sulfuric acid, SAJ first grade, ≥95.0%
Sigma-Aldrich
Potassium thiocyanate, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Sulfuric acid solution, 0.1 M
Sigma-Aldrich
Benzaldehyde, SAJ special grade, ≥98.0%
Sigma-Aldrich
Sulfuric acid solution, 0.05 M
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Ethyl hexanoate, SAJ special grade, ≥99.0%
Sigma-Aldrich
Sulfuric acid solution, 1.5 M
Sigma-Aldrich
Octanoic acid, ≥96.0%
Sigma-Aldrich
Sulfuric acid, JIS special grade, ≥95.0%
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Sulfuric acid solution, 0.01 M
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 23 ga
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 23 ga, for use with autosampler
Sigma-Aldrich
2-Decanone, ≥98%
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), metal alloy fiber, df 100 μm(PDMS, needle size 23 ga, for use with autosampler
Sigma-Aldrich
2,3-Pentanedione, natural, ≥96%, FG
Sigma-Aldrich
Skatole, ≥98%
Sigma-Aldrich
4-Ethylguaiacol, natural, FG
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sigma-Aldrich
Isovaleraldehyde, 97%
Sigma-Aldrich
Ethyl butyrate, ≥98%, FCC, FG