Skip to Content
Merck
All Photos(1)

Key Documents

24-5710

Sigma-Aldrich

Potassium thiocyanate

JIS special grade, ≥99.5%

Synonym(s):

Potassium rhodanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KSCN
CAS Number:
Molecular Weight:
97.18
Beilstein:
3594799
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
≥99.5%
grade:
JIS special grade
form:
solid

grade

JIS special grade

Assay

≥99.5%

form

solid

availability

available only in Japan

pH

5.3-8.7 (25 °C, 97.2 g/L)

mp

173 °C (lit.)

SMILES string

[K]SC#N

InChI

1S/CHNS.K/c2-1-3;/h3H;/q;+1/p-1

InChI key

ZNNZYHKDIALBAK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongtao Bian et al.
The journal of physical chemistry. B, 116(49), 14426-14432 (2012-11-28)
Microscopic structures and dynamics of aqueous salt solutions were investigated with the ultrafast vibrational energy exchange method and anisotropy measurements. In KSCN aqueous solutions of various concentrations, the rotational time constants of SCN(-) anions are proportional to the viscosities of
Altamir Benedito de Sousa et al.
Reproductive toxicology (Elmsford, N.Y.), 23(4), 568-577 (2007-03-06)
A study was performed at term and at weaning to verify the toxic effects of the prenatal exposure to potassium cyanide (KCN) and potassium thiocyanate (KSCN) in pregnant Wistar rats. Females received daily in drinking water the doses: 1, 3
Meiyun Zhang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 75(3), 1013-1017 (2010-01-12)
A highly sensitive visible spectrophotometric method has been developed to determine ethamsylate in this paper, which is based on using Cu(II) as spectroscopic probe reagent. The study indicates that in the presence of SCN(-) and KNO(3), Cu(II) is reduced to
A Botti et al.
The journal of physical chemistry. B, 113(29), 10014-10021 (2009-07-03)
The results of a neutron diffraction experiment performed on aqueous solutions of KSCN are analyzed and discussed in comparison with similar data for KCl and KOH solutions. The effect of the different ions on the structure of water and the
Fei Wang et al.
The Journal of organic chemistry, 77(8), 4148-4151 (2012-03-27)
A concise approach to various benzisothiazol-3(2H)-one derivatives has been developed by copper-catalyzed the reaction of o-bromobenzamide derivatives with potassium thiocyanate (KSCN) in water. The reaction proceeds via a tandem reaction with S-C bond and S-N bond formation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service