Merck
  • GADD34 inhibits activation-induced apoptosis of macrophages through enhancement of autophagy.
GADD34 inhibits activation-induced apoptosis of macrophages through enhancement of autophagy.
Scientific reports (2015-02-11)
Sachiko Ito, Yuriko Tanaka, Reina Oshino, Keiko Aiba, Suganya Thanasegaran, Naomi Nishio, Ken-ichi Isobe
ABSTRACT
Autophagy is a common physiological function in all eukaryotes. The process is induced by depletion of nutrients including amino acids. GADD34 is expressed following DNA damage, ER stresses and amino acid deprivation. Here, we investigated the effects of GADD34 on autophagy and cell activation in macrophages. The deprivation of tyrosine and cysteine markedly induced the expression of GADD34 in macrophages. LPS stimulation combined with tyrosine/cysteine-deprivation initially activated macrophages, but then shifted to cell death in late phase of stimulation. When LPS stimulation was combined with tyrosine/cysteine-deprivation, a deficiency of GADD34 enhanced cell activation signaling such as Src-family, Erk1/2, p38 MAPK and Akt. In the late phase of stimulation, a deficiency of GADD34 increased apoptosis more than that in wild-type macrophages. Further we found that mTOR-S6K signaling was highly enhanced in GADD34-deficient macrophages compared with wild-type cells when cells were treated by LPS combined with tyrosine/cysteine-deprivation. LC3-II was increased by LPS stimulation combined with tyrosine/cysteine-deprivation. Defective GADD34 reduced LC3-II and autophagosome formation induced by LPS-stimulation and tyrosine/cysteine-deprivation compared with that seen in wild-type macrophages. These results indicates that GADD34 enhances autophagy and suppresses apoptosis stimulated by LPS combined with amino acid deprivation through regulation of mTOR signaling pathway in macrophages.
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by phenol extraction
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.