Skip to Content
Merck
All Photos(3)

Key Documents

S7653

Sigma-Aldrich

Sodium chloride

BioXtra, ≥99.5% (AT)

Synonym(s):

Natrium Chloride, Sodium monochloride, Halite, NaCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.25
grade:
for analytical purposes
form:
powder
solubility:
H2O: 1 M, clear, colorless
Pricing and availability is not currently available.

grade

for analytical purposes

Quality Level

product line

BioXtra

Assay

≥99.5% (AT)

form

powder

technique(s)

immunoblotting: suitable
immunoprecipitation (IP): suitable

impurities

filtertest, complies
insoluble matter, complies

pH

5.0-8.0 (20 °C, 1 M in H2O)

mp

801 °C (lit.)

solubility

H2O: 1 M, clear, colorless

density

2.16 g/cm3 at 25 °C (77 °F)

anion traces

bromide (Br-): ≤0.01%
iodide (I-): ≤0.001%
phosphate (PO43-): ≤0.0005%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
As: ≤0.0001%
Ba: ≤0.0005%
Bi: ≤0.0005%
Ca: ≤0.002%
Cd: ≤0.0005%
Co: ≤0.0005%
Cr: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0001%
K: ≤0.005%
Li: ≤0.0005%
Mg: ≤0.0005%
Mn: ≤0.0005%
Mo: ≤0.0005%
Ni: ≤0.0005%
Pb: ≤0.0005%
Sr: ≤0.0005%
Zn: ≤0.0005%

absorption

≤0.01 at 260
≤0.01 at 280 in H2O at 1 M

λ

(260 nm Amax: ≤ 0.01)

SMILES string

[Na+].[Cl-]

application(s)

genome editing
microbiology

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium chloride, also known as halite salt, is one of nature′s most common minerals. It is a known electrolyte that maintains cellular integrity and homeostasis by controlling osmotic pressure and water balance. Sodium chloride is widely used in the food industry as food additive and preservative due to its antimicrobial property.[1] It is added to laboratory buffers to increase their ionic strength.[2] NaCl also finds application in the extraction of DNA. Sodium and Chloride are major electrolytes in biological functions.

Application

Sodium chloride has been used to:
  • Prepare PBS (Phosphate-buffered saline) solution.[3][4][5]
  • in pre-treatment of tracheal cartilage tissue samples before decellularization.[6]
  • As a component of trypticase soy agar medium to culture bacteria. [7]
  • Prepare lysis buffer.[5]
It is also used for regulation of acid-base equilibrium of body fluids along with sodium bicarbonate.

Features and Benefits

Tested for trace elemental impurities to ensure high purity.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influence of the ionic strength on the polyelectrolyte multilayers' permeability
Antipov et al.
Langmuir, 19, 2444-2448 (2003)
Salt in food processing; usage and reduction: a review
Albarracin, Willian et al
International Journal of Food Science and Technology, 46, 1329-1336 (2011)
Alain Boldini et al.
Scientific reports, 9(1), 11989-11989 (2019-08-21)
A variety of modeling frameworks have been proposed for ionic polymer metal composites (IPMCs), but the physical underpinnings of their actuation remain elusive. A critical step toward the validation of existing theories and transition to engineering practice entails the design
An optimized method for measurement of gamma-H2AX in blood mononuclear and cultured cells.
Muslimovic A, et al.
Nature Protocols, 3(7), 1187-1193 (2008)
Beneficial effects of four Bacillus strains on the larval cultivation of Litopenaeus vannamei
Luis-Villase et al.
Aquaculture (Amsterdam, Netherlands), 321, 136-144 (2011)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service