Merck
  • The activity of silver nanoparticles (Axonnite) on clinical and environmental strains of Acinetobacter spp.
The activity of silver nanoparticles (Axonnite) on clinical and environmental strains of Acinetobacter spp.
Burns : journal of the International Society for Burn Injuries (2014-08-26)
Monika E Łysakowska, Anna Ciebiada-Adamiec, Leszek Klimek, Monika Sienkiewicz
ABSTRACT
Acinetobacter baumannii isolates are responsible for a high number of wound infections. The reason of this study was to evaluate the activity of silver nanoparticles obtained by microexplosion against wide range of Acinetobacter spp. Susceptibility to silver nanoparticles was tested by microdilution method, susceptibility to antibiotics was evaluated by the disc-diffusion method. All strains of Acinetobacter spp. were sensitive to AgNPs at low concentrations. The values of the MIC for strains of Acinetobacter spp. were 0.39 and 0.78μg/mL. In general, strains inhibited by 0.78μg/mL of AgNPs were more resistant to antibiotics than Acinetobacter strains for which MIC=0.39μg/mL (p=0.023). The AgNPs in Axonnite seems to be a good alternative for other antimicrobials to treat wound infections caused by multidrug resistant Acinetobacter spp. strains because of its high activity.
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
USP
Amikacin, United States Pharmacopeia (USP) Reference Standard
Amikacin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR