Merck
  • Time-dependent effects of hydrophobic amine-containing drugs on lysosome structure and biogenesis in cultured human fibroblasts.
Time-dependent effects of hydrophobic amine-containing drugs on lysosome structure and biogenesis in cultured human fibroblasts.
Journal of pharmaceutical sciences (2014-07-22)
Randall Logan, Alex C Kong, Jeffrey P Krise
ABSTRACT
Many weakly basic amine-containing drugs are known to be extensively sequestered in acidic lysosomes by an ion trapping-type mechanism. The entrapment of drugs in lysosomes has been shown to influence drug activity, cancer cell selectivity, and pharmacokinetics and can cause the hyperaccumulation of various lipids associated with lysosomes. In this work, we have investigated the prolonged time-dependent effects of drugs on lysosomal properties. We have evaluated two amine-containing drugs with intermediate (propranolol) and high (halofantrine) relative degrees of lipophilicity. Interestingly, the cellular accumulation kinetics of these drugs exhibited a biphasic characteristic at therapeutically relevant exposure levels with an initial apparent steady-state occurring at 2 days followed by a second stage of enhanced accumulation. We provide evidence that this secondary drug accumulation coincides with the nuclear localization of transcription factor EB, a master regulator of lysosome biogenesis, and the appearance of an increased number of smaller and lipid-laden lysosomes. Collectively, these results show that hydrophobic lysosomotropic drugs can induce their own cellular accumulation in a time-dependent fashion and that this is associated with an expanded lysosomal volume. These results have important therapeutic implications and may help to explain sources of variability in drug pharmacokinetic distribution and elimination properties observed in vivo.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium pyruvate, SAJ special grade, ≥95.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
SAFC
HEPES
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%