Skip to Content
Merck
All Photos(1)

Key Documents

54457

Sigma-Aldrich

HEPES

≥99.5% (titration), powder, for molecular biology, BioUltra

Synonym(s):

4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H18N2O4S
CAS Number:
Molecular Weight:
238.30
Beilstein:
883043
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

Product Name

HEPES, BioUltra, for molecular biology, ≥99.5% (T)

grade

Molecular Biology
for molecular biology

product line

BioUltra

Assay

≥99.5% (T)

form

powder

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

ign. residue

≤0.1% (as SO4)

loss

≤0.5% loss on drying, 110 °C

pH

5.0-6.5 (25 °C, 1 M in H2O)

useful pH range

6.8-8.2

pKa (25 °C)

7.5

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.045
λ: 280 nm Amax: 0.035

SMILES string

OCCN1CCN(CC1)CCS(O)(=O)=O

InChI

1S/C8H18N2O4S/c11-7-5-9-1-3-10(4-2-9)6-8-15(12,13)14/h11H,1-8H2,(H,12,13,14)

InChI key

JKMHFZQWWAIEOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

HEPES has been described as one of the best all-purpose buffers available for biological research. At biological pH, the molecule is zwitterionic, and is effective as a buffer at pH 6.8 to 8.2. HEPES has been used in a wide variety of applications, including tissue culture. It is commonly used to buffer cell culture media in air. HEPES finds its usage in in vitro experiments on Mg.

Application

HEPES has been used as a component of:
  • DMEM (Dulbecco′s modified Eagle′s medium) used for the cultivation of CHO-GPR40?cells
  • RNA buffer (for gel-filtration chromatography) and transcription buffer (for transcription reaction)
  • Recapture solution to re-capture RNA exosomes from the glycerol gradient
  • ExoI and ExoII extraction solution as well as light and heavy gradient solution for gradient centrifugation
  • 2X HBS buffer that is used in transfection process for the production of recombinant lentivectors

Other Notes

3′-Terminal labeling of RNA with T4 RNA ligasecitation; Isolation and analysis of nuclear RNA - component of reaction buffer, prehybridization buffer, hybridization buffercitation.
Easily compare specifications for HEPES products with the HEPES specification table.
3′-Terminal labeling of RNA with T4 RNA ligase; Isolation and analysis of nuclear RNA - component of reaction buffer, prehybridization buffer, hybridization buffer.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Optogenetic interrogation of neural circuits: technology for probing mammalian brain structures.
Zhang F, et al.
Nature Protocols, 5(3), 439-439 (2010)
The correction of the metabolic parameters of msg-induced obesity in rats by 2-[4-(benzyloxy) phenoxy] acetic acid.
Konopelniuk V, et al.
Journal of Nutrition and Intermediary Metabolism, 13, 1-9 (2018)
F. Cobianchi et al.
Methods in Enzymology, 152, 103-103 (1987)
Affinity Purification of the RNA Degradation Complex, the Exosome, from HEK-293 Cells.
Domanski M and LaCava J
Bio-protocol, 7(8) (2017)
Sample Preparation of Telomerase Subunits for Crystallization.
Huang J, et al.
Bio-protocol, 5(16) (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service