Merck
  • Electrochemical in situ fabrication of titanium dioxide-nanosheets on a titanium wire as a novel coating for selective solid-phase microextraction.
Electrochemical in situ fabrication of titanium dioxide-nanosheets on a titanium wire as a novel coating for selective solid-phase microextraction.
Journal of chromatography. A (2014-07-20)
Yi Li, Min Zhang, Yaoxia Yang, Xuemei Wang, Xinzhen Du
ABSTRACT
A novel TiO2-nanosheets coated fiber for solid-phase microextraction (SPME) was fabricated by anodization of Ti wire substrates in ethylene glycol with concentrated NH4F. The in situ fabricated TiO2-nanosheets were densely embedded into Ti substrates with about 1μm long, 300nm wide and 80nm thick. The as-fabricated TiO2-nanosheets coating was employed to extract polycyclic aromatic hydrocarbons, phthalates and ultraviolet (UV) filters in combination with high performance liquid chromatography-UV detection (HPLC-UV). It was found that the TiO2-nanosheets coating exhibited high extraction capability and good selectivity for some UV filters frequently used in cosmetic sunscreen formulations. The main parameters affecting extraction performance were investigated and optimized. Under the optimized conditions, the calibration graphs were linear in the range of 0.1-400μgL(-1). The limits of detection of the proposed method were between 0.026μgL(-1) and 0.089μgL(-1) (S/N=3). The single fiber repeatability varied from 4.50% to 8.76% and the fiber-to-fiber reproducibility ranged from 7.75% to 9.64% for the extraction of spiked water with 50μgL(-1) UV filters (n=5). The SPME-HPLC-UV method was successfully established for the selective preconcentration and sensitive detection of target UV filters from real environmental water samples. Recovery of UV filters spiked at 10μgL(-1) and 25μgL(-1) ranged from 88.8% to 107% and the relative standard deviations were less than 9.8%. Furthermore the in situ growth of the TiO2-nanosheets coating was performed in a highly reproducible manner and the TiO2-nanosheets coated fiber has high mechanical strength, good stability and long service life.
Product Description
Supelco
Octisalate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Phenanthrene, analytical standard, for environmental analysis
Supelco
Fluoranthene, analytical standard
Supelco
Melting point standard 79-81°C, analytical standard
Sigma-Aldrich
2-Ethylhexyl salicylate, ≥99%
Sigma-Aldrich
Fluoranthene, 98%
Sigma-Aldrich
Ethylene glycol 5 M solution
Sigma-Aldrich
Phenanthrene, sublimed grade, ≥99.5%
Supelco
Phenanthrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
2-Ethylhexyl salicylate, 99%
Sigma-Aldrich
Phenanthrene, 98%
USP
Octisalate, United States Pharmacopeia (USP) Reference Standard
Supelco
2-Ethylhexyl salicylate, analytical standard
Sigma-Aldrich
Anthracene, sublimed grade, ≥99%
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylene glycol, SAJ first grade, ≥99.0%
Sigma-Aldrich
Ethylene glycol, JIS special grade, ≥99.5%
Supelco
Pyrene, analytical standard
Supelco
Anthracene, analytical standard
Pyrene, BCR®, certified reference material
Supelco
Ethylene glycol, analytical standard
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
USP
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyrene, puriss. p.a., for fluorescence, ≥99.0% (GC)
Sigma-Aldrich
Pyrene, sublimed grade, 99%
Sigma-Aldrich
Pyrene, 98%
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%
Supelco
Naphthalene, analytical standard
Sigma-Aldrich
Naphthalene, 99%