Skip to Content
Merck
All Photos(2)

Documents

293237

Sigma-Aldrich

Ethylene glycol

spectrophotometric grade, ≥99%

Synonym(s):

1,2-Ethanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2OH
CAS Number:
Molecular Weight:
62.07
Beilstein:
505945
EC Number:
MDL number:
UNSPSC Code:
12352137
eCl@ss:
39020501
PubChem Substance ID:
NACRES:
NA.21

grade

spectrophotometric grade

vapor density

2.1 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)

Assay

≥99%

form

liquid

autoignition temp.

752 °F

expl. lim.

15.3 %

technique(s)

UV/Vis spectroscopy: suitable

impurities

<0.050% water

refractive index

n20/D 1.431 (lit.)

bp

195-198 °C

mp

−13 °C (lit.)

density

1.113 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 210 nm Amax: 1.00
λ: 220 nm Amax: 0.40
λ: 250 nm Amax: 0.10
λ: 300 nm Amax: 0.01

SMILES string

OCCO

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethylene glycol solvent is featured in:
  • Tandem affinity purification-mass spectrometry (TAP-MS) characterization of protein complexes
  • Measurement of total protein content using a ninhydrin-based quantification method

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

239.0 °F - open cup

Flash Point(C)

115 °C - open cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Boris N Khlebtsov et al.
Langmuir : the ACS journal of surfaces and colloids, 28(24), 8994-9002 (2012-03-13)
We describe a novel strategy for the fabrication of plasmonic nanopowders (dried gold nanoparticles) by using wet chemical nanoparticle synthesis, PEG-SH functionalization, and a standard freeze-drying technique. Our strategy is illustrated by successful fabrication of different plasmonic nanopowders, including gold
Study of polyol synthesis reaction parameters controlling high yield of silver nanocubes.
Panfilova EV, et al.
Colloid Journal, 74(1), 99-109 (2012)
Synthesis and characterization of yttrium iron garnet nanoparticles.
Vaqueiro P, et al.
Journal of Solid State Chemistry, 126(2), 161-168 (1996)
Lennart Treuel et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 11(14), 3093-3099 (2010-09-04)
Interactions between differently functionalised silver and gold nanoparticles (NPs) as well as polystyrene nanoparticles with bovine serum albumin (BSA) are studied using circular dichroism (CD) spectroscopy. It is found that the addition of NPs to the protein solution destroys part
Anomalously increased effective thermal conductivities of ethylene glycol-based nanofluids containing copper nanoparticles.
Eastman JA, et al.
Applied Physics Letters, 78(6), 718-720 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service