Skip to Content
Merck
All Photos(1)

Documents

BCR177R

Pyrene

BCR®, certified reference material

Synonym(s):

Benzo[def]phenanthrene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10
CAS Number:
Molecular Weight:
202.25
Beilstein:
1307225
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

145-148 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

c1cc2ccc3cccc4ccc(c1)c2c34

InChI

1S/C16H10/c1-3-11-7-9-13-5-2-6-14-10-8-12(4-1)15(11)16(13)14/h1-10H

InChI key

BBEAQIROQSPTKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyrene belongs to polycyclic aromatic hydrocarbon group. These are organic compounds which have two or more fused benzene rings in linear, angular and cluster arrangements. Pyrene is a four ring PAH.

Analysis Note

For more information please see:
BCR177R

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

435.2 °F

Flash Point(C)

224 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomohiro Kato et al.
Journal of the American Chemical Society, 135(2), 741-750 (2012-12-18)
Although distance dependence of Förster resonance energy transfer (FRET) is well-studied and FRET has been extensively applied as "molecular ruler", only limited examples of orientation-dependent FRET have been reported. To create a robust FRET system that precisely reflects the orientation
Intramolecular pyrene excimer fluorescence: a probe of proximity and protein conformational change.
S S Lehrer
Methods in enzymology, 278, 286-295 (1997-01-01)
Michael E Østergaard et al.
Chemical Society reviews, 40(12), 5771-5788 (2011-04-14)
Pyrene-functionalized oligonucleotides (PFOs) are increasingly explored as tools in fundamental research, diagnostics and nanotechnology. Their popularity is linked to the ability of pyrenes to function as polarity-sensitive and quenchable fluorophores, excimer-generating units, aromatic stacking moieties and nucleic acid duplex intercalators.
Pyrene-tagged dendritic catalysts noncovalently grafted onto magnetic Co/C nanoparticles: an efficient and recyclable system for drug synthesis.
Michel Keller et al.
Angewandte Chemie (International ed. in English), 52(13), 3626-3629 (2013-02-22)
Yanna Liang
Pyrene Degradation by Mycobacterium Sp Kms: Biochemical Pathway, Enzymatic Mechanisms, and Humic Acid Effect, 1- 4 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service