Merck
  • Chemical composition and functional properties of native chestnut starch (Castanea sativa Mill).
Chemical composition and functional properties of native chestnut starch (Castanea sativa Mill).
Carbohydrate polymers (2013-04-03)
Bruno R Cruz, Ana S Abraão, André M Lemos, Fernando M Nunes
ABSTRACT
Starch isolation methods can change their physico-chemical and functional characteristics hindering the establishment of a starch-food functionality relation. A simple high yield and soft isolation method was applied for chestnut (Castanea sativa Mill) starch consisting in steeping and fruit disintegration in a 25 mM sodium bisulfite solution and purification by sedimentation. Starch integrity, physico-chemical composition, morphology and functional properties were determined, being observed significant differences from previous described methods for chestnut starch isolation. The X-ray pattern was of B-type, with a degree of crystallinity ranging from 51% to 9%, dependent on the starch moisture content. The onset, peak, and conclusion gelatinization temperatures were 57.1°C, 61.9°C and 67.9°C, respectively. Total amylose content was 26.6%, and there was not found any evidence for lipid complexed amylose. Swelling power at 90°C was 19 g/g starch, and the amount of leached amylose was 78% of the total amylose content. Native chestnut starch presents a type B pasting profile similar to corn starch but with a lower gelatinization (56.1°C) and peak viscosity (79.5°C) temperatures, making native chestnut starch a potential technological alternative to corn starch, especially in application where lower processing temperatures are needed.
Sigma-Aldrich
Starch from wheat, Unmodified
Sigma-Aldrich
Starch from potato, suitable for electrophoresis
Sigma-Aldrich
Amylose from potato, used as amylase substrate
Sigma-Aldrich
Starch from potato, Powder
Sigma-Aldrich
Starch from potato, Soluble
Sigma-Aldrich
Starch, from potato, tested according to Ph. Eur.
Sigma-Aldrich
Starch, puriss. p.a., from potato, reag. ISO, reag. Ph. Eur., soluble
Sigma-Aldrich
Starch from corn, practical grade
Sigma-Aldrich
Starch from corn
Supelco
Starch from corn, for use with Total Dietary Fiber Control Kit, TDF-C10
Supelco
Starch from corn, analytical standard, analytical standard for Starch Assay Kits SA-20 and STA-20
Sigma-Aldrich
Starch from rice
Sigma-Aldrich
Starch from corn