Skip to Content
Merck
All Photos(4)

Key Documents

S4180

Sigma-Aldrich

Starch from corn

practical grade

Synonym(s):

Corn starch, Starch from maize

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

biological source

corn

Quality Level

grade

practical grade

form

solid

color

white

useful pH range

4.3 - 6.7

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Starch is a natural storage carbohydrate[1] polymer produced by plants[2] and varies in its form and function.[1] It is considered as a macroconstituent of food[1] and serves as an energy source. It is renewable and can be biodegraded.[2] Glucose polymers, amylose and amylopectin forms starch molecule.[1] Starch influences the food structure and quality.[3] It is a native high amylose corn starch.

Application

Starch from corn has been used:
  • in the preparation of test diet pellets[4]
  • to study the activity of recombinant β-amylase (BAM) proteins[5]
  • to study the amylopectin molecular structures and functional properties[6]

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Form and functionality of starch
Copeland L, et al.
Food Hydrocolloids, 23(6), 1527-1534 (2009)
Catalytically-inactive beta-amylase BAM4 required for starch breakdown in Arabidopsis leaves is a starch-binding-protein
Li J, et al.
Archives of Biochemistry and Biophysics, 489(1-2), 92-98 (2009)
Aspects of the physical chemistry of starch
Parker R and Ring SG
Journal of Cereal Science, 34(1), 1-17 (2001)
Relationships between amylopectin molecular structures and functional properties of different-sized fractions of normal and high-amylose maize starches
Lin L, et al.
Food Hydrocolloids, 52, 359-368 (2016)
Starch nanoparticles: a review
Le Corre D, et al.
Biomacromolecules, 11(5), 1139-1153 (2010)

Questions

1–4 of 4 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. What is the average starch content of this product?

    1 answer
    1. Starch is composed primarily of amylose and amylopectin. This product is not tested to determine the content of either component or total starch content. Historical information indicates this product is high in amylose, at approximately 70%. In addition, this material may have up to 13% moisture content. Please see the link below to review a sample or lot specific Certificate of Analysis:
      https://www.sigmaaldrich.com/product/sial/s4180#product-documentation

      Helpful?

  4. What is the amylose percentage in the product? Thank you!

    1 answer
    1. The amount of amylose has not been determined and likely varies from lot to lot.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service