Skip to Content
Merck
All Photos(1)

Documents

S5296

Supelco

Starch from corn

analytical standard, analytical standard for Starch Assay Kits SA-20 and STA-20

Synonym(s):

Corn starch, Starch from maize

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

biological source

corn

Quality Level

grade

analytical standard

analyte chemical class(es)

oligosaccharides

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicholas Arpaia et al.
Nature, 504(7480), 451-455 (2013-11-15)
Intestinal microbes provide multicellular hosts with nutrients and confer resistance to infection. The delicate balance between pro- and anti-inflammatory mechanisms, essential for gut immune homeostasis, is affected by the composition of the commensal microbial community. Regulatory T cells (Treg cells)
Daniëlle Haenen et al.
The Journal of nutrition, 143(12), 1889-1898 (2013-10-18)
Consumption of resistant starch (RS) has been associated with various intestinal health benefits, but knowledge of its effects on global gene expression in the colon is limited. The main objective of the current study was to identify genes affected by
Karolien Decamps et al.
Food chemistry, 148, 235-239 (2013-11-23)
The impact of pyranose oxidase (P₂O), glucose oxidase (GO) and H₂O₂ on gluten agglomeration during wheat flour gluten-starch separation was studied. Analysis of gluten aggregate sizes in batter formed from wheat flour dough revealed that increasing levels of oxidising agents
Shujun Wang et al.
Food & function, 4(11), 1564-1580 (2013-10-08)
Starch is the most important glycemic carbohydrate in foods. The relationship between the rate and extent of starch digestion to produce glucose for absorption into the bloodstream and risk factors for diet-related diseases is of considerable nutritional interest. Native starch
E van der Hoeven-Hangoor et al.
Poultry science, 92(10), 2713-2723 (2013-09-21)
Microbiota plays a role in the release and absorption of nutrients from feed components, thereby affecting digesta composition and moisture content of the excreta. The objective of the current study was to determine the effects of 5 different diets varying

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service