Merck
  • Deficits of peripheral olfactory inputs reduce cell proliferation in the adult subventricular and subgranular zones.
Deficits of peripheral olfactory inputs reduce cell proliferation in the adult subventricular and subgranular zones.
Neuroscience letters (2013-03-05)
Zhixiang Xu, Yunling Gao, Fuqiang Xu
ABSTRACT
Constitutive adult neurogenesis mainly occurs in the subgranular zone (SGZ) and subventricular zone (SVZ) of the mammalian brain. Intrinsic and extrinsic factors regulate adult neurogenesis processes including cell proliferation, migration, integration and neural survival. The potential contribution of olfactory sensory input to the regulation of adult neurogenesis, especially neurogenesis in the dentate gyrus, however, is not well studied. In the present study, we examined the effects of deficits in the peripheral olfactory inputs on cell proliferation in the adult SGZ and SVZ. With an anosmic model produced by ZnSO4 irrigation of the olfactory epithelium, we found that in the adult SVZ and SGZ the numbers of both BrdU labeled cells and doublecortin labeled cells (immature neurons) were significantly decreased, whereas the number of adult stem cells was not significantly altered. These results suggested that olfactory sensory input may play roles in regulating adult neurogenesis.
Sigma-Aldrich
Zinc sulfate heptahydrate, ≥99.95% trace metals basis
Sigma-Aldrich
Zinc sulfate heptahydrate, BioUltra, for molecular biology, 2.0 M in H2O
Sigma-Aldrich
Zinc sulfate heptahydrate, suitable for plant cell culture
Sigma-Aldrich
Zinc sulfate heptahydrate, meets USP testing specifications
Sigma-Aldrich
Zinc sulfate heptahydrate, BioReagent, suitable for cell culture
Sigma-Aldrich
Zinc sulfate heptahydrate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Zinc sulfate heptahydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5%
Sigma-Aldrich
Zinc sulfate heptahydrate, ACS reagent, 99%
Sigma-Aldrich
Zinc sulfate heptahydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Zinc sulfate solution, 0.3 N