Skip to Content
Merck
All Photos(4)

Documents

Z4750

Sigma-Aldrich

Zinc sulfate heptahydrate

ReagentPlus®, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnSO4 · 7H2O
CAS Number:
Molecular Weight:
287.56
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55

Agency

suitable for SM 5210

Quality Level

product line

ReagentPlus®

Assay

≥99.0%

form

powder

SMILES string

O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

InChI

1S/H2O4S.7H2O.Zn/c1-5(2,3)4;;;;;;;;/h(H2,1,2,3,4);7*1H2;/q;;;;;;;;+2/p-2

InChI key

RZLVQBNCHSJZPX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Zinc sulfate heptahydrate has been used as:
  • A reagent in the isolation of genomic DNA from eukaryotic green alga named Chlamydomonas reinhardtii.
  • A component of a cell culture media (Jensen′s chemically defined media).
  • A reactant to prepare zinc thiourea sulfate salts as NLO materials.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of Pseudomonas aeruginosa c-di-GMP High and Low Subpopulations Using Flow-assisted Cell Sorting (FACS) and Quantitative Reverse Transcriptase PCR (qRT-PCR)
Armbruster CR and Parsek MR
Bio-protocol, 11, e3891-e3891 (2021)
Isolation of Genomic DNA from Chlamydomonas reinhardtii
Wittkopp TM
Bio-protocol, e2838-e2838 (2018)
Growth of zinc thiourea sulfate (ZTS) single crystals: a potential semiorganic NLO material
Ushasree PM, et al.
Journal of Crystal Growth, 197, 216-220 (1999)
Dervis Özcelik et al.
Biological trace element research, 150(1-3), 342-349 (2012-10-12)
Zinc is an element that under physiological conditions preferentially binds to and is a potent inducer of metallothionein under physiological conditions. The present study was conducted to explore whether zinc supplementation morphologically and biochemically protects against diabetic nephropathy through modulation
O I Smian et al.
Likars'ka sprava, (3-4)(3-4), 67-71 (2013-01-30)
The article analyzed 60 patients with community-aquired pneumonia from ages 6 to 18 years. The dependences of trace levels of zinc and copper in the serum of these children on the activity of the inflammatory process. Analysis of survey results

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service