Skip to Content
Merck
All Photos(3)

Documents

Z0251

Sigma-Aldrich

Zinc sulfate heptahydrate

BioReagent, suitable for cell culture

Synonym(s):

Zinc Sulfate hydrate, Zinc sulphate heptahydrate, ZnSO4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnSO4 · 7H2O
CAS Number:
Molecular Weight:
287.56
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.71

product line

BioReagent

Assay

>99.0%

technique(s)

cell culture | mammalian: suitable

solubility

H2O: 100 mg/mL, clear, colorless

SMILES string

O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

InChI

1S/H2O4S.7H2O.Zn/c1-5(2,3)4;;;;;;;;/h(H2,1,2,3,4);7*1H2;/q;;;;;;;;+2/p-2

InChI key

RZLVQBNCHSJZPX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaohua Huang et al.
Nutrients, 9(10) (2017-10-24)
The trace element zinc plays an important role in human life. Zinc deficiency impairs growth, reproduction, metabolism and immunity in both human and animals. Thus, zinc supplementation is recommended in daily life. However, the effect of long-term chronic zinc supplementation
Hitoshi Tsujimoto et al.
Frontiers in physiology, 9, 380-380 (2018-05-01)
Mosquito-transmitted viral pathogens, such as dengue and Zika, afflict tens of thousands of people every year. These viruses are transmitted during the blood-feeding process that is required for mosquito reproduction, the most important vector being Aedes aegypti. While vertebrate blood
Brandon Mapes et al.
Physiological genomics, 46(8), 302-308 (2014-02-20)
1α,25-Dihydroxyvitamin D3 [1α,25(OH)2D3] is a steroid hormone derived from circulating 25(OH) vitamin D [25(OH)D] with chemopreventive effects in colorectal cancer. 1α,25(OH)2D3 acts through transcriptional mechanisms; however, our understanding of vitamin D transcriptional responses in the colon is derived from studies
Keizo Tokuhiro et al.
Developmental cell, 46(5), 627-640 (2018-08-21)
The zona pellucida surrounding ovulated eggs regulates monospermic fertilization necessary for successful development. Using mouse transgenesis, we document that the N terminus of ZP2 is sufficient for sperm binding to the zona matrix and for in vivo fertility. Sperm binding is
Nicole A J Krentz et al.
Developmental cell, 41(2), 129-142 (2017-04-26)
During pancreatic development, proliferating pancreatic progenitors activate the proendocrine transcription factor neurogenin 3 (NEUROG3), exit the cell cycle, and differentiate into islet cells. The mechanisms that direct robust NEUROG3 expression within a subset of progenitor cells control the size of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service