Merck
Optimization of biodiesel production by supercritical methyl acetate.
Bioresource technology (2013-01-24)
Fadjar Goembira, Shiro Saka
ABSTRACT
This work has been done to find out the optimum condition of supercritical methyl acetate method in biodiesel production. The reaction temperature, pressure, time and molar ratio in methyl acetate to oil were the key parameters that must all be considered to produce an optimum condition. Evaluation of thermal decomposition on products, cis-trans isomerization and tocopherol content were required to further optimize the reaction condition. It was, therefore, concluded that for the supercritical methyl acetate method, reaction condition of 350 °C/20 MPa/45 min/42 M ratio gave the highest yields of FAME (96.7 wt.%) and triacetin (8.8 wt.%). Yet, at such a reaction condition, the optimum reaction condition was compromised due particularly to the unavoidable thermal decomposition of products, and tocopherols as natural anti-oxidants.
Sigma-Aldrich
Methyl acetate, ≥98%, FG
Sigma-Aldrich
Methyl acetate, natural, 98%, FG
Supelco
Methyl acetate, analytical standard
Sigma-Aldrich
Methyl acetate, anhydrous, 99.5%
Sigma-Aldrich
Methyl acetate, JIS special grade, ≥99.5%
Sigma-Aldrich
Methyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Methyl acetate, ReagentPlus®, 99%