Skip to Content
Merck
All Photos(2)

Key Documents

186325

Sigma-Aldrich

Methyl acetate

ReagentPlus®, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH3
CAS Number:
Molecular Weight:
74.08
Beilstein:
1736662
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.21

vapor density

2.55 (vs air)

Quality Level

vapor pressure

165 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

99%

form

liquid

autoignition temp.

936 °F

expl. lim.

16 %

refractive index

n20/D 1.361 (lit.)

bp

57-58 °C (lit.)

mp

−98 °C (lit.)

density

0.934 g/mL at 25 °C

SMILES string

COC(C)=O

InChI

1S/C3H6O2/c1-3(4)5-2/h1-2H3

InChI key

KXKVLQRXCPHEJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Its IR spectra in the vapor phase and in solution form (in CS2 and CCl4) have been reported. It can be synthesized from dimethyl ether via carbonylation in the presence of halide-free catalysts based on zeolites. It has also been reported to be formed during the synthesis of poly(vinyl) alcohol (PVA). It undergoes transesterification reaction with n-octanol in the presence of Amberlyst 15 catalyst to afford octyl acetate and methanol.
Methyl acetate is an aliphatic ester that can be prepared via carbonylation of dimethyl ether over zeolites. MA is formed as a by-product during the preparation of polyvinyl alcohol from acetic acid and methanol.

Application

Methyl acetate may be used for the preparation of fatty acid methyl esters and triacetin from rapeseed oil via non-catalytic trans-esterification reaction under super-critical conditions.
Methyl acetate may be used in the following:
  • As acyl acceptor in the preparation of biodiesel.
  • Synthesis of ethanol.
  • Preparation of n-butyl acetate, via transesterification reaction with n-butanol in the presence of acidic catalysts.
It may also be used as a precursor in the synthesis of the following:
  • acetic anhydride
  • methyl acrylate
  • vinyl acetate
  • ethyl amide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

8.6 °F - closed cup

Flash Point(C)

-13 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haipeng Lu et al.
Journal of the American Chemical Society, 141(12), 4919-4927 (2019-03-02)
The marriage of colloidal semiconductor nanocrystals and functional organic molecules has brought unique opportunities in emerging photonic and optoelectronic applications. Traditional semiconductor nanocrystals have been widely demonstrated to initiate efficient triplet energy transfer at the nanocrystal-acene interface. Herein, we report
Dimethyl ether carbonylation to methyl acetate over HZSM-35.
Liu J, et al.
Catalysis Letters, 139(1-2), 33-37 (2010)
Study on the kinetics of enzymatic interesterification of triglycerides for biodiesel production with methyl acetate as the acyl acceptor.
Xu Y, et al.
Journal of Molecular Catalysis. B, Enzymatic, 32(5), 241-245 (2005)
Catalysts, Kinetics, and Reactive Distillation for Methyl Acetate Synthesis.
Zuo C, et al.
Industrial & Engineering Chemistry Research, 53(26), 10540-10548 (2014)
Selective carbonylation of dimethyl ether to methyl acetate catalyzed by acidic zeolites.
Patricia Cheung et al.
Angewandte Chemie (International ed. in English), 45(10), 1617-1620 (2006-01-31)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service