Skip to Content
Merck
All Photos(1)

Key Documents

W267619

Sigma-Aldrich

Methyl acetate

natural, 98%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH3
CAS Number:
Molecular Weight:
74.08
FEMA Number:
2676
Beilstein:
1736662
EC Number:
Council of Europe no.:
213c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.023
NACRES:
NA.21

grade

FG
Halal
Kosher
natural

Quality Level

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 175.105

vapor density

2.55 (vs air)

vapor pressure

165 mmHg ( 20 °C)

Assay

98%

form

liquid

autoignition temp.

936 °F

expl. lim.

16 %

refractive index

n20/D 1.361 (lit.)

bp

57-58 °C (lit.)

mp

−98 °C (lit.)

density

0.934 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

ethereal; fruity; sweet

SMILES string

COC(C)=O

InChI

1S/C3H6O2/c1-3(4)5-2/h1-2H3

InChI key

KXKVLQRXCPHEJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

8.6 °F - closed cup

Flash Point(C)

-13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Franc Avbelj et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(9), 3137-3141 (2009-02-10)
Recent calorimetric measurements of the solvation enthalpies of some dipeptide analogs confirm our earlier prediction that the principle of group additivity is not valid for the interaction of the peptide group with water. We examine the consequences for understanding the
Marco Candelaresi et al.
The journal of physical chemistry. A, 113(46), 12783-12790 (2009-10-02)
The solvation dynamics of methyl acetate in heavy water are analyzed by means of two-dimensional infrared spectroscopy, in conjunction with Car-Parrinello molecular dynamics simulations. The C horizontal lineO stretching infrared band of methyl acetate in water splits into a doublet
Sara M Aschmann et al.
Environmental science & technology, 45(16), 6896-6901 (2011-07-22)
3-Methoxy-3-methyl-1-butanol [CH(3)OC(CH(3))(2)CH(2)CH(2)OH] is used as a solvent for paints, inks, and fragrances and as a raw material for the production of industrial detergents. A rate constant of (1.64 ± 0.18) × 10(-11) cm(3) molecule(-1) s(-1) for the reaction of 3-methoxy-3-methyl-1-butanol
Frank Kjeldsen et al.
Journal of the American Society for Mass Spectrometry, 22(8), 1441-1452 (2011-09-29)
Probing the mechanism of electron capture dissociation on variously modified model peptide polycations has resulted in discovering many ways to prevent or reduce N-Cα bond fragmentation. Here we report on a rare finding of how to increase the backbone bond
Fadjar Goembira et al.
Bioresource technology, 131, 47-52 (2013-01-24)
This work has been done to find out the optimum condition of supercritical methyl acetate method in biodiesel production. The reaction temperature, pressure, time and molar ratio in methyl acetate to oil were the key parameters that must all be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service