Merck
  • BNIP3-dependent mitophagy promotes cytosolic localization of LC3B and metabolic homeostasis in the liver.
BNIP3-dependent mitophagy promotes cytosolic localization of LC3B and metabolic homeostasis in the liver.
Autophagy (2021-01-19)
Maya Z Springer, Logan P Poole, Lauren E Drake, Althea Bock-Hughes, Michelle L Boland, Alexandra G Smith, John Hart, Aparajita H Chourasia, Ivan Liu, Grazyna Bozek, Kay F Macleod
ABSTRACT
Mitophagy formed the basis of the original description of autophagy by Christian de Duve when he demonstrated that GCG (glucagon) induced macroautophagic/autophagic turnover of mitochondria in the liver. However, the molecular basis of liver-specific activation of mitophagy by GCG, or its significance for metabolic stress responses in the liver is not understood. Here we show that BNIP3 is required for GCG-induced mitophagy in the liver through interaction with processed LC3B; an interaction that is also necessary to localize LC3B out of the nucleus to cytosolic mitophagosomes in response to nutrient deprivation. Loss of BNIP3-dependent mitophagy caused excess mitochondria to accumulate in the liver, disrupting metabolic zonation within the liver parenchyma, with expansion of zone 1 metabolism at the expense of zone 3 metabolism. These results identify BNIP3 as a regulator of metabolic homeostasis in the liver through its effect on mitophagy and mitochondrial mass distribution.Abbreviations: ASS1, arginosuccinate synthetase; BNIP3, BCL2/adenovirus E1B interacting protein 3; CV, central vein; GCG - glucagon; GLUL, glutamate- ammonia ligase (glutamine synthetase); HCQ, hydroxychloroquine; LIR, LC3-interacting region; MAP1LC3B/LC3B, microtubule-associated protein 1 light chain 3 beta; mtDNA:nucDNA, ratio of mitochondrial DNA to nuclear DNA; PV, periportal vein; TOMM20, translocase of outer mitochondrial membrane protein 20.
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, suitable for (for molecular biology), Non-acetylated
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Leupeptin, microbial, ≥90% (HPLC)
Sigma-Aldrich
Anti-p21/WAF1/Cip1 Antibody, clone CP74, clone CP74, Upstate®, from mouse
Sigma-Aldrich
Oligomycin, A mixture of A, B, and C isomers.
Sigma-Aldrich
Anti-BNIP3 antibody, Mouse monoclonal, clone ANa40, purified from hybridoma cell culture
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Anti-BNIP3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Aprotinin Nicotiana tobacco, >= 5TIU/mg protein, >= 98 % SDS-PAGE | 9087-70-1, recombinant, expressed in Nicotiana (tobacco), ≥5 TIU/mg protein, ≥98% (SDS-PAGE)