Skip to Content
Merck
All Photos(2)

Key Documents

05-655

Sigma-Aldrich

Anti-p21/WAF1/Cip1 Antibody, clone CP74

clone CP74, Upstate®, from mouse

Synonym(s):

Anti-CAP20, Anti-CIP1, Anti-MDA-6, Anti-P21, Anti-SDI1, Anti-WAF1, Anti-p21CIP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

CP74, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CDKN1A(1026)

Specificity

Cip1

Immunogen

Recombinant full-length human Cip1 with an N-terminal histidine tag; epitope corresponds to amino acids 1-80 of full-length human Cip1

Application

Anti-p21/WAF1/Cip1 Antibody, clone CP74 is a high quality Mouse Monoclonal Antibody for the detection of p21/WAF1/Cip1 & has been validated in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

routinely evaluated by immunoblot on nuclear extract from HeLa cells

Target description

21kDa

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Format: Purified
Protein G Purified

Storage and Stability

2 years at -20°C

Analysis Note

Control
Positive Antigen Control: Catalog #12-309, Hela cell nuclear extract. Add an equal volume of Laemmli reducing sample buffer to 10 μL of extract and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced extract per lane for minigels.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongye Huang et al.
Biomedical reports, 7(1), 36-40 (2017-07-08)
Wnt signaling is critical in embryonic development and post-embryonic tissue homeostasis. The aim of the present study was to evaluate the expression levels of canonical Wnt signaling genes in porcine somatic cell nuclear transfer (SCNT) embryos. Quantitative polymerase chain reaction
N-methyl-N'-nitro-N-nitrosoguanidine (MNNG) triggers MSH2 and Cdt2 protein-dependent degradation of the cell cycle and mismatch repair (MMR) inhibitor protein p21Waf1/Cip1.
Jascur, T; Fotedar, R; Greene, S; Hotchkiss, E; Boland, CR
The Journal of Biological Chemistry null
STX140 and STX641 cause apoptosis via the intrinsic mitochondrial pathway and down-regulate survivin and XIAP expression in ovarian and prostate cancer cells.
Foster PA, Ho YT, Newman SP, Leese MP, Potter BV, Reed MJ, Purohit A
Anticancer research null
Z Y Yang et al.
Nature medicine, 1(10), 1052-1056 (1995-10-01)
The p21 gene encodes a cyclin-dependent kinase inhibitor that affects cell-cycle progression, but the potential of this gene product to serve as a tumour suppressor in vivo has not been established. In this report, we show that the growth of
Heat shock protein 90 inhibitors attenuate LPS-induced endothelial hyperpermeability.
Chatterjee, A; Snead, C; Yetik-Anacak, G; Antonova, G; Zeng, J; Catravas, JD
American Journal of Physiology. Lung Cellular and Molecular Physiology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service